Skip to Content
Merck
All Photos(1)

Key Documents

58980C

SAFC

Diamond Vitamin Tween® 80 Solution 40x

frozen liquid, suitable for cell culture

Pharma Manufacturing

Synonym(s):

BME Vitamins 100x solution

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352207
NACRES:
NA.75

description

for research or for further manufacturing use

Quality Level

sterility

sterile; sterile-filtered

form

frozen liquid

technique(s)

cell culture | mammalian: suitable

color

faint yellow to dark yellow

pH

3.5-6.5

shipped in

dry ice

storage temp.

−20°C

Looking for similar products? Visit Product Comparison Guide

General description

Diamond Vitamin Tween® 80 Solution 40X supplement provides nutrients and reduces the biosynthetic burden of cells in vitro. Vitamin supplements enhance the growth and prolong cell viability in cell culture media.

Application

Diamond Vitamin Tween® 80 Solution 40x has been used as a supplement in TYI-S33 medium to grow Entamoeba histolytica trophozoites.
Diamond Vitamin Tween® 80 Solution 40x has been used as a supplement:
  • to culture Spironucleus salmonicida
  • to culture Giardia intestinalis
  • to culture Entamoeba histolytica trophozoites

Legal Information

TWEEN is a registered trademark of Croda International PLC

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

143.6 °F

Flash Point(C)

62 °C


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

[Percutaneous puncture of the femoral vein for hemodialysis].
B Grabensee et al.
Deutsche medizinische Wochenschrift (1946), 102(39), 1401-1401 (1977-09-30)
Effect of Clinoptilolite and Sepiolite Nanoclays on Human and Parasitic Highly Phagocytic Cells
Yanis Toledano-Maga?a
BioMed Research International (2015)
Jan Šlapeta et al.
Parasitology, 143(8), 1012-1025 (2016-03-24)
Establishing a health screening protocol is fundamental for successful captive breeding and release of wildlife. The aim of this study was to undertake a parasitological survey focusing on the presence of trypanosomes in a cohort of Regent Honeyeaters, Anthochaera phrygia
Jon Jerlström-Hultqvist et al.
Eukaryotic cell, 11(11), 1353-1361 (2012-09-18)
Eukaryotic microbes are highly diverse, and many lineages remain poorly studied. One such lineage, the diplomonads, a group of binucleate heterotrophic flagellates, has been studied mainly due to the impact of Giardia intestinalis, an intestinal, diarrhea-causing parasite in humans and
Ranae M Ranade et al.
Antimicrobial agents and chemotherapy, 59(11), 7128-7131 (2015-09-02)
The methionyl-tRNA synthetase (MetRS) is a novel drug target for the protozoan pathogen Giardia intestinalis. This protist contains a single MetRS that is distinct from the human cytoplasmic MetRS. A panel of MetRS inhibitors was tested against recombinant Giardia MetRS

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service