Skip to Content
Merck
All Photos(1)

Key Documents

511-500

Sigma-Aldrich

Bronchia/Trachea Epithelial Cell Growth Medium (500 ml)

Synonym(s):

Primary Cell Culture Media

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352207
NACRES:
NA.71
Pricing and availability is not currently available.

form

liquid

Quality Level

shelf life

6 mo.

manufacturer/tradename

Cell Applications, Inc

shipped in

wet ice

storage temp.

2-8°C

General description

All-in-one ready-to-use media
Bronchia/Trachea Epithelial Cell Growth Medium is an all-in-one ready-to-use media. It fully supports cell growth on its own.

Application

Bronchia/Trachea Epithelial Cell Growth Medium (500 ml) has been used to culture lung spheroid cells(LSCs)[1] and human Het1A cells.[2]

Quality

Each lot was tested for the ability to support plating, spreading and proliferation

Preparation Note

contained in protocol

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Maria Davern et al.
Translational oncology, 14(6), 101062-101062 (2021-03-26)
Use of immune checkpoint inhibitors (ICIs) with chemotherapy to enhance responses in oesophageal adenocarcinoma (OAC) is an attractive approach. We identified subpopulations of OAC cells expressing inhibitory immune checkpoint (IC) ligands (PD-L1, PD-L2 and CD160) and receptors (PD-1, TIGIT, TIM-3
Phuong-Uyen C Dinh et al.
Respiratory research, 18(1), 132-132 (2017-07-02)
Resident stem and progenitor cells have been identified in the lung over the last decade, but isolation and culture of these cells remains a challenge. Thus, although these lung stem and progenitor cells provide an ideal source for stem-cell based

Protocols

Technical information for working with human bronchial epithelial cell line including thawing, subculturing and cryopreservation.

Technical information for working with human bronchial epithelial cell line including thawing, subculturing and cryopreservation.

Technical information for working with human bronchial epithelial cell line including thawing, subculturing and cryopreservation.

Technical information for working with human bronchial epithelial cell line including thawing, subculturing and cryopreservation.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service