Skip to Content
Merck
All Photos(1)

Documents

48795

Sigma-Aldrich

Manganese(II) chloride 0.1 M solution

Synonym(s):

Manganese(II) chloride solution, Additive Screening Solution 26/Kit-No 78374

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
MnCl2
CAS Number:
Molecular Weight:
125.84
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.32

density

1.01 g/mL at 20 °C

storage temp.

2-8°C

SMILES string

Cl[Mn]Cl

InChI

1S/2ClH.Mn/h2*1H;/q;;+2/p-2

InChI key

GLFNIEUTAYBVOC-UHFFFAOYSA-L

Looking for similar products? Visit Product Comparison Guide

Application

Manganese(II) chloride 0.1 M solution has been used:
  • as a component of reprogramming media for culturing mouse neonatal tail-tip fibroblasts
  • in the mutagenesis of WT clone by error-prone polymerase chain reaction (PCR)
  • as a standard in isothermal titration calorimetry and differential scanning calorimetry

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Robert W Siegel
Methods in molecular biology (Clifton, N.J.), 504, 351-383 (2009-01-23)
Many biosensors depend on molecular recognition reagents to achieve highly sensitive and specific detection levels of an analyte of interest. Although new and improved detection platforms continue to be developed, improvements in the affinity and specificity of the molecular recognition
Alexey Gribenko et al.
Journal of molecular biology, 425(18), 3429-3445 (2013-07-06)
MntC is a metal-binding protein component of the Mn²⁺-specific mntABC transporter from the pathogen Staphylococcus aureus. The protein is expressed during the early stages of infection and was proven to be effective at reducing both S. aureus and Staphylococcus epidermidis
Xiaowen Wang et al.
Scientific reports, 6, 23017-23017 (2016-03-16)
We have recently shown that a combination of microRNAs, miR combo, can directly reprogram cardiac fibroblasts into functional cardiomyocytes in vitro and in vivo. However, direct reprogramming strategies are inefficient and slow. Moving towards the eventual goal of clinical application
Tora Sund Morken et al.
Pediatric research, 73(2), 171-179 (2012-11-24)
Hypoxia-ischemia (HI) induces delayed inflammation and long-term gray and white matter brain injury that may be altered by hyperoxia. HI and 2 h of hyperoxia (100% O2) or room air (21% O2) in 7-d-old (P7) rats were studied by magnetic resonance
Oliviero L Gobbo et al.
Contrast media & molecular imaging, 7(6), 542-546 (2012-09-20)
Lung parenchyma remains one of the most difficult tissues to be imaged by means of magnetic resonance imaging (MRI). Several MRI techniques are routinely used for lung imaging. However, manganese-enhancement MRI (MEMRI) technique has not been associated with pulmonary MRI.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service