Skip to Content
Merck
All Photos(1)

Key Documents

47612

Sigma-Aldrich

Folinic acid calcium salt hydrate

BioXtra, ≥99.0% (HPLC)

Synonym(s):

5-Formyl-5,6,7,8-tetrahydrofolic acid calcium salt, 5-Formyl-5,6,7,8-tetrahydropteroyl-L-glutamic acid calcium salt, 5-HCO-H4PteGlu, Calcium folinate, Citrovorum factor calcium salt, Leucovorin calcium salt

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C20H21CaN7O7 · xH2O
CAS Number:
Molecular Weight:
511.50 (anhydrous basis)
Beilstein:
5723924
MDL number:
UNSPSC Code:
12352106
PubChem Substance ID:
NACRES:
NA.28

biological source

synthetic

Quality Level

product line

BioXtra

Assay

≥99.0% (HPLC)

form

powder or crystals

technique(s)

cell culture | mammalian: suitable

color

white to faint yellow

mp

240-250 °C

application(s)

cell analysis

SMILES string

[Ca++].[H]O[H].[H]C(=O)N1C(CNc2ccc(cc2)C(=O)N[C@@H](CCC([O-])=O)C([O-])=O)CNc3nc(N)nc(O)c13

InChI

1S/C20H23N7O7.Ca/c21-20-25-16-15(18(32)26-20)27(9-28)12(8-23-16)7-22-11-3-1-10(2-4-11)17(31)24-13(19(33)34)5-6-14(29)30;/h1-4,9,12-13,22H,5-8H2,(H,24,31)(H,29,30)(H,33,34)(H4,21,23,25,26,32);/q;+2/p-2/t12?,13-;/m0./s1

InChI key

KVUAALJSMIVURS-ZEDZUCNESA-L

Looking for similar products? Visit Product Comparison Guide

General description

Calcium salt of the (6R,S)-mixture of diastereomers of 5-formyl-5,6,7,8-tetrahydrofolate.
Folinic acid, also called as calcium leucovorin, is usually available as an amorphous powder and lacks odor. This activated form of vitamin B9 is off-white to light beige in color. Folic acid (FA) is a water-soluble vitamin. Folinic acid is the 5-formyl derivative of tetrahydrofolic acid and is the reduced form of folic acid.

Application

Calcium folinate offsets the effects of folic acid antagonists that act by binding dihydrofolate reductase and blocking the conversion of folic acid to tetrahydrofolate. In medicine, calcium folinate is used after treatment with methotrexate to reduce the toxicity of the methotrexate. Calcium folinate is also used to enhance the cytotoxic effect of 5-fluorouracil in the treatment of cancer.
Folinic acid calcium salt hydrate has been used in the preparation of topical 1% and 2.5% folinic acid creams for use in wound healing studies. It has also been used as a component in the chemically defined-Chinese hamster ovary medium (CD-CHO) to culture CHO DUKX B11.

Biochem/physiol Actions

Folinic acid serves as a cofactor and participates in one-carbon-transfer reactions. It helps in improving wound healing in rats. Folinic acid plays a major role in children with autism to enhance verbal communication.

Other Notes

Review

Pictograms

Health hazardExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Resp. Sens. 1 - Skin Irrit. 2 - Skin Sens. 1 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

L.O. Pont et al.
Anal. Profiles Drug Subst., 8, 315-315 (1979)
Nutritional Regulation of Mitochondrial Function
Mitochondria, 93-126 (2019)
OPP Labeling Enables Total Protein Synthesis Quantification in CHO Production Cell Lines at the Single-Cell Level
Nagelreiter F, et al.
Biotechnology Journal, 13(4), 1700492-1700492 (2018)
Calcium Leucovorin
Analytical Profiles of Drug Substances, 8, 315-350 (1979)
Yoon Jeong et al.
Journal of clinical medicine, 9(8) (2020-08-23)
Methotrexate is widely used as an intraocular chemotherapy for vitreoretinal lymphoma (VRL). Although corneal toxicity has been reported in patients after intravitreal methotrexate injections, the incidence, outcome, and mechanism of the toxicity are unclear. Herein, we performed a clinical study

Articles

Cancer research innovations address the complexity of the disease, providing advanced technologies for varied applications.

Cancer research innovations address the complexity of the disease, providing advanced technologies for varied applications.

Biofiles reviews innovative technologies for cancer research, reflecting the complexity of the disease.

Cancer research innovations address the complexity of the disease, providing advanced technologies for varied applications.

See All

Chromatograms

application for HPLCapplication for HPLC

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service