Skip to Content
Merck
All Photos(1)

Key Documents

43725

Supelco

Delphinidin chloride

analytical standard

Synonym(s):

3,3′,4′,5,5′,7-Hexahydroxyflavylium chloride, 3,5,7-Trihydroxy-2-(3,4,5-trihydroxyphenyl)-1-benzopyrylium chloride

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C15H11ClO7
CAS Number:
Molecular Weight:
338.70
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

Assay

≥95.0% (HPLC)

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

food and beverages

format

neat

storage temp.

−20°C

SMILES string

[Cl-].Oc1cc(O)c2cc(O)c([o+]c2c1)-c3cc(O)c(O)c(O)c3

InChI

1S/C15H10O7.ClH/c16-7-3-9(17)8-5-12(20)15(22-13(8)4-7)6-1-10(18)14(21)11(19)2-6;/h1-5H,(H5-,16,17,18,19,20,21);1H

InChI key

FFNDMZIBVDSQFI-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Delphinidin chloride is commonly found in delphinium flowers. It can be obtained, via hydrolysis of delphinin chloride.

Application

Delphinidin chloride is an anthocyanin standard, which has been used in the identification and quantification of crude anthocyanin extract from Hibiscus sabdariffa, using high performance liquid chromatography coupled with diode array detector (HPLC-DAD).
Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Biochem/physiol Actions

Anthocyanidin. Antioxidant. Pigment in grapes, cranberries, and pomegranates.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

A J Kortstee et al.
Transgenic research, 20(6), 1253-1264 (2011-02-23)
A mutant allele of the transcription factor gene MYB10 from apple induces anthocyanin production throughout the plant. This gene, including its upstream promoter, gene coding region and terminator sequence, was introduced into apple, strawberry and potato plants to determine whether
Yoosoo Yang et al.
Proceedings of the National Academy of Sciences of the United States of America, 107(51), 22145-22150 (2010-12-08)
Neuronal SNARE proteins mediate neurotransmitter release at the synapse by facilitating the fusion of vesicles to the presynaptic plasma membrane. Cognate v-SNAREs and t-SNAREs from the vesicle and the plasma membrane, respectively, zip up and bring about the apposition of
Hirofumi Inoue et al.
Nutrition research (New York, N.Y.), 32(5), 357-364 (2012-06-02)
Delphinidin-3-O-galactoside (D3G) is a water-soluble anthocyanin with antioxidant activity. (-)-Epigallocatechin-3-gallate (EGCG) is also known as a powerful antioxidant but concomitantly possesses a prooxidative property. We hypothesized that D3G is capable of protecting the EGCG-induced cytotoxicity and endoplasmic reticulum (ER) stress
Natural Products, Volume 2, 2 (2006)
Hina Kausar et al.
Cancer letters, 325(1), 54-62 (2012-06-05)
Berry anthocyanidins (cyanidin, malvidin, peonidin, petunidin and delphinidin) have increasingly been explored for their anticancer effects; however, their combinatorial effects as a mixture, as present in blueberry, bilberry and Indian blackberry ('Jamun') remain untested. In this study, we demonstrate for

Articles

Antioxidants protect biological systems from oxidative damage produced by oxygen-containing free radicals and from redoxactive transition metal ions such as iron, copper, and cadmium.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service