Skip to Content
Merck
All Photos(1)

Key Documents

43659

Sigma-Aldrich

Glucosylsphingosine

≥98.0% (TLC)

Synonym(s):

(2S,3R,4E)-2-Amino-3-hydroxy-4-octadecen-1-yl β-D-glucopyranoside, 1-β-D-Glucosylsphingosine, Glucosyl-C18-sphingosine

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C24H47NO7
CAS Number:
Molecular Weight:
461.63
Beilstein:
4333674
MDL number:
UNSPSC Code:
12352211
PubChem Substance ID:
NACRES:
NA.85

Quality Level

Assay

≥98.0% (TLC)

form

powder

lipid type

sphingolipids

storage temp.

−20°C

SMILES string

O[C@@H]1[C@@H](O)[C@H](OC[C@H](N)[C@H](O)/C=C/CCCCCCCCCCCCC)O[C@H](CO)[C@H]1O

InChI

1S/C24H47NO7/c1-2-3-4-5-6-7-8-9-10-11-12-13-14-15-19(27)18(25)17-31-24-23(30)22(29)21(28)20(16-26)32-24/h14-15,18-24,26-30H,2-13,16-17,25H2,1H3/b15-14+/t18-,19+,20?,21+,22+,23?,24+/m0/s1

InChI key

HHJTWTPUPVQKNA-JLRUQHRASA-N

Biochem/physiol Actions

Glucosylsphingosine is a cytotoxic compound. Accumulation of glucosylsphingosine in brain and other tissues occurs in patients with Gaucher disease, which is an inherited deficiency of lysosomal glucocerebrosidase, which converts glucosylsphingosine to glucose and sphingosine.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Lulu Kang et al.
Journal of human genetics, 62(8), 763-768 (2017-03-31)
Gaucher disease (GD) is an inherited metabolic disorder that involves accumulation of glycolipid glucocerebroside in monocyte-macrophage cells, which can result in multiple organ damage. Enzyme replacement and substrate reduction therapies have improved the potential for early diagnosis and treatment. Determining
N G Conradi et al.
Acta neuropathologica, 75(4), 385-390 (1988-01-01)
Splenectomy in children with the Norrbottnian type of Gaucher disease is followed by increased blood levels of glucosylceramide and impaired neurological and mental status. High blood levels are associated with an increased accumulation of glucosylceramide in perivascular Gaucher cells in
Shiny Nair et al.
Molecular genetics and metabolism, 129(4), 286-291 (2020-02-12)
In Gaucher disease type 1 (GD1), genetic deficiency of lysosomal glucocerebrosidase results in the accumulation of glucosylceramide and glucosylsphingosine (GlcSph), that underlie chronic lipid-mediated metabolic inflammation. An important age-related phenotype is high risk of monoclonal gammopathy (MG), including multiple myeloma.
E Beutler
Blood reviews, 2(1), 59-70 (1988-03-01)
Gaucher disease is a glycolipid storage disorder characterized by accumulation of glucocerebroside in the liver, spleen, and bones, and caused by a deficiency of glucocerebrosidase. Glucocerebrosidase cDNA has been cloned and sequenced, and much has been learned about the synthesis
A Kaloterakis et al.
Journal of internal medicine, 246(6), 587-590 (2000-01-05)
Chronic Gaucher disease [GD] in association with systemic AL amyloidosis is extremely rare. We describe a 46-year-old Greek male with chronic GD confirmed by low glucocerebroside activity in fibroblasts and N370S/L444P mutations at the cerebrosidase gene, who also had systemic

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service