Skip to Content
Merck
All Photos(2)

Key Documents

2852

Sigma-Aldrich

Acid Hematoxylin Solution

Acid Hematoxylin Solution
Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352106
NACRES:
NA.47
Pricing and availability is not currently available.

form

solution

shelf life

Expiry date on the label.

IVD

for in vitro diagnostic use

concentration

1 g/L

pH

3.3

application(s)

hematology
histology

storage temp.

room temp

Application

Nuclear counterstain. Included as a component in the Sigma Fetal Hemoglobin Procedure 285.

Components

1 g/L certified hematoxylin with aluminum sulfate, sodium iodate and stabilizers. pH 3.3

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Nicholas J Reichart et al.
The ISME journal, 14(11), 2851-2861 (2020-09-06)
Metagenomic studies have revolutionized our understanding of the metabolic potential of uncultured microorganisms in various ecosystems. However, many of these genomic predictions have yet to be experimentally tested, and the functional expression of genomic potential often remains unaddressed. In order
Jiang Jiang et al.
Pharmaceutical research, 32(9), 2851-2862 (2015-03-17)
This study seeks to develop fiber membranes for local sustained delivery of 25-hydroxyvitamin D3 to induce the expression and secretion of LL-37 at or near the surgical site, which provides a novel therapeutic approach to minimize the risk of infections.

Questions

Reviews

Active Filters

  1. 1 Ratings-Only Review

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service