Skip to Content
Merck
All Photos(1)

Key Documents

17794

Sigma-Aldrich

Isorhamnetin

≥95.0% (HPLC)

Synonym(s):

3′-Methoxy-3,4′,5,7-tetrahydroxyflavone, 3′-Methylquercetin, 3,4′,5,7-Tetrahydroxy 3′-methoxyflavone, Isorhamnetol, Quercetin 3′-methyl ether

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C16H12O7
CAS Number:
Molecular Weight:
316.26
Beilstein:
44723
EC Number:
MDL number:
UNSPSC Code:
12352205
PubChem Substance ID:
NACRES:
NA.47

Assay

≥95.0% (HPLC)

application(s)

metabolomics
vitamins, nutraceuticals, and natural products

SMILES string

COc1cc(ccc1O)C2=C(O)C(=O)c3c(O)cc(O)cc3O2

InChI

1S/C16H12O7/c1-22-11-4-7(2-3-9(11)18)16-15(21)14(20)13-10(19)5-8(17)6-12(13)23-16/h2-6,17-19,21H,1H3

InChI key

IZQSVPBOUDKVDZ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Isorhamnetin is an O-methylated flavonol found in fruits, plants, and medicinal herbs. It is a metabolite of quercetin.

Application

Isorhamnetin (IRN) has been used:
  • as a neuroprotective agent to test its effect on scopolamine-induced cortico-hippocampal learning and memory deficiency in mice
  • as a reference standard in quadrupole time-of-flight mass spectrometry (QTOF MS) to analyze and quantify the phenolic compounds present in honey extract
  • as a reference standard to determine the phenolic profile of Artemisia species using Reversed-phase high-performance liquid chromatography with diode array detection and electrospray ionization mass spectrometry (RP-HPLC-DAD-ESI-TQ-MS/MS)

Biochem/physiol Actions

Isorhamnetin exerts antioxidant, anti-inflammatory, anti-cancer, anti-bacterial, and anti-viral effects. It has shown to be a potential neuroprotective agent by inhibiting neurodegeneration in the hippocampus under Diabetes mellitus (DM) conditions. Isorhamnetin at low levels may serve as an anti-diabetic agent by promoting glucose uptake in skeletal muscle cells and maintaining glucose homeostasis in hyperglycemic conditions. It regulates cell signaling pathways such as Nuclear factor kappa B (NF-κB), phosphatidylinositol 3-kinase (PI3K)/protein kinase B (AKT), mitogen-activated protein kinase (MAPK) etc. Isorhamnetin elicits protective and therapeutic effects on atherosclerosis and cardiovascular diseases.
Isorhamnetin inhibits adipogenesis by interfering with differentiation of adipose stem cells, by a mechanism involving stabilization of β-catenin and up-regulating the Wnt signaling pathway.

Packaging

Bottomless glass bottle. Contents are inside inserted fused cone.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Gang Gong et al.
Biomedicine & pharmacotherapy = Biomedecine & pharmacotherapie, 128, 110301-110301 (2020-06-06)
Isorhamnetin is one of the most important active ingredients in the fruits of Hippophae rhamnoides L. and the leaves of Ginkgo biloba L., which possesses extensive pharmacological activities. At present, there have been numerous investigations on isorhamnetin, which has the
Quercetin and its metabolite isorhamnetin promote glucose uptake through different signalling pathways in myotubes
Jiang H, et al.,
Scientific Reports, 1-15 (2019)
Felipe Surco-Laos et al.
Food & function, 2(8), 445-456 (2011-07-22)
Quercetin is a major flavonoid in the human diet and the most commonly used in studies of biological activity. Most of the knowledge about its biological effects has originated from in vitro studies while in vivo data are scarce. Quercetin
Nida Jamali-Raeufy et al.
Journal of chemical neuroanatomy, 102, 101709-101709 (2019-11-08)
Isorhamnetin, a derivative of quercetin, exerts antioxidant and anti-inflammatory effects in different diseases, and we examined its protective effects against diabetes-related changes in the brain. A single dose of a freshly prepared solution of streptozotocin (STZ) (60 mg/kg body weight) was
Lutz Franke et al.
Journal of medicinal chemistry, 50(11), 2640-2646 (2007-04-28)
A natural product collection and natural-product-derived combinatorial libraries were virtually screened for potential inhibitors of human 5-lipoxygenase (5-LO) activity. We followed a sequential ligand-based approach in two steps. First, similarity searching with a topological pharmacophore descriptor (CATS 2D method) was

Articles

Antioxidants protect biological systems from oxidative damage produced by oxygen-containing free radicals and from redoxactive transition metal ions such as iron, copper, and cadmium.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service