Skip to Content
Merck
All Photos(1)

Key Documents

17783

Sigma-Aldrich

Calcein-AM

BioReagent, suitable for fluorescence, ≥90% (HPLC)

Synonym(s):

Calcein acetoxymethyl ester, Calcein O,O′-diacetate tetrakis(acetoxymethyl) ester

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C46H46N2O23
CAS Number:
Molecular Weight:
994.86
MDL number:
UNSPSC Code:
12171500
PubChem Substance ID:
NACRES:
NA.32
Pricing and availability is not currently available.

product line

BioReagent

Assay

≥90% (HPLC)

fluorescence

λex 496 nm; λem 516 nm in 0.1 M Tris pH 8.0, esterase; Ca2+

suitability

suitable for fluorescence

storage temp.

−20°C

SMILES string

CC(=O)OCOC(=O)CN(CC(=O)OCOC(C)=O)Cc1c(OC(C)=O)ccc2c1Oc3c(CN(CC(=O)OCOC(C)=O)CC(=O)OCOC(C)=O)c(OC(C)=O)ccc3C24OC(=O)c5ccccc45

InChI

1S/C46H46N2O23/c1-25(49)60-21-64-39(55)17-47(18-40(56)65-22-61-26(2)50)15-32-37(68-29(5)53)13-11-35-43(32)70-44-33(16-48(19-41(57)66-23-62-27(3)51)20-42(58)67-24-63-28(4)52)38(69-30(6)54)14-12-36(44)46(35)34-10-8-7-9-31(34)45(59)71-46/h7-14H,15-24H2,1-6H3

InChI key

XKFSBWQWNMZWFA-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Liposomes contain a self-quenching fluorescent dye called calcein. It is a fluorescent dye with excitation and emission wavelengths of 495/515 nm, respectively. It self-quenches at concentrations above 70mM and is commonly used as an indicator of lipid vesicle leakage. The leakage rate is calculated on the basis of the fluorescence increment as the entrapped calcein is leaked out of the liposomal compartment. It is used as a complexometric indicator for titration of calcium ions with EDTA, and for fluorometric determination of calcium.[1][2]

Application

Calcein-AM has been used in flow cytometry.[3]
Non-fluorescent cell permeable derivative of calcein, becomes fluorescent on hydrolysis. Used as a neutral substrate for multidrug resistance protein, MRP; employed in tumor research.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Y S Tsao et al.
Biochemistry, 24(5), 1092-1098 (1985-02-26)
Sendai virus induced liposome leakage has been studied by using liposomes containing a self-quenching fluorescent dye, calcein. The liposomes used in this study were prepared by a freeze and thaw method and were composed of phosphatidylcholine, phosphatidylserine, and phosphatidylethanolamine (1:2.60:1.48
T M Allen et al.
Biochimica et biophysica acta, 597(2), 418-426 (1980-04-10)
Efflux of contents from small unilamellar vesicles of various compositions, conaining a highly quenched fluorescent compound (calcein, 175 mM) was determined as a function of temperature in the presence and absence of human serum. Efflux of calcein from the liposomes
Xiao-Cong Huang et al.
Biochemical pharmacology, 85(9), 1257-1268 (2013-02-19)
High intrinsic or acquired expression of membrane spanning, adenosine triphosphate binding cassette (ABC) transporter proteins, such as P-glycoprotein (P-gp), in cancers represents a major impediment to chemotherapy, with accelerated drug efflux leading to multi-drug resistance (MDR). Although ABC transporter inhibitors
Bao-Ling Du et al.
Journal of biomedical materials research. Part A, 103(4), 1533-1545 (2014-07-22)
Biological materials combined with genetically-modified neural stem cells (NSCs) are candidate therapy targeting spinal cord injury (SCI). Based on our previous studies, here we performed gelatin sponge (GS) scaffold seeded with neurotrophin-3 (NT-3) and its receptor TrkC gene modifying NSCs
Johan van der Stok et al.
Tissue engineering. Part A, 21(9-10), 1495-1506 (2015-01-30)
A promising bone graft substitute is porous titanium. Porous titanium, produced by selective laser melting (SLM), can be made as a completely open porous and load-bearing scaffold that facilitates bone regeneration through osteoconduction. In this study, the bone regenerative capacity

Articles

Sigma-Aldrich presents a Biofiles on Detect, Visualize and Quantify Single Post-Translational Modifications

Cell based assays for cell proliferation (BrdU, MTT, WST1), cell viability and cytotoxicity experiments for applications in cancer, neuroscience and stem cell research.

Cell based assays for cell proliferation (BrdU, MTT, WST1), cell viability and cytotoxicity experiments for applications in cancer, neuroscience and stem cell research.

Cell based assays for cell proliferation (BrdU, MTT, WST1), cell viability and cytotoxicity experiments for applications in cancer, neuroscience and stem cell research.

See All

Related Content

Step-by-step protocol for generating apical-out human gut organoids for microbiome, ADME/Tox, viral and gastrointestinal related disease research. See the complete organoid culture protocol.

Questions

1–8 of 8 Questions  
  1. What live cell dye (DNA non-binding) is recommended for staining live cells while ensuring that the cells remain viable after staining, particularly for bacterial cells? Would using DSMO be suitable, or is there an alternative stain recommendation?

    1 answer
    1. Calcein AM may not be soluble in water. DMSO can be used as the solvent and there is no mention of solubility in water or alcohol. The free acid is soluble in water and ethanol. However, only DMSO is listed for the solubility of Calcein AM. Concerns about DMSO affecting bacteria viability may lead to a switch from the hexa ester to the free acid form. Maintaining a 1:500 dilution of DMSO in the staining solution is advisable to avoid potential viability issues.

      Helpful?

  2. Recommend fluorescent dyes for live bacterial cell sorting, especially those validated that the dye enters in bacteria and are not transferred to daughter cells.

    1 answer
    1. Calcein-AM is a fluorescent dye used to stain live bacterial cells for flow cytometry application. Product 17783, Calcein-AM, is suitable for fluorescence and is at least 96.0% HPLC pure. For dyes that can stain bacteria but do not enter bacterial cells, you may find the products SCT126, BioTracker 510 Green C4(FM1-43) Synaptic Dye, and SCT127, BioTracker 640 Red C2(FM4-64) Synaptic Dye may be suitable.

      Helpful?

  3. What is the appearance of Calcein-AM, Product No. 17783?

    1 answer
    1. Product No. 17783 can be a film, oil, paste or solid.

      Helpful?

  4. What are the spectral properties of Calcein-AM, Product 17783?

    1 answer
    1. The emission maximum for Calcein-AM is 516 nm, with excitation at 496 nm in 0.1M Tris pH 8.0 (following hydrolysis by esterase enzyme, bound to Ca(2+)).

      Helpful?

  5. What is the Department of Transportation shipping information for this product?

    1 answer
    1. Transportation information can be found in Section 14 of the product's (M)SDS.To access the shipping information for this material, use the link on the product detail page for the product.

      Helpful?

  6. How is Calcein-AM, Product 17783, typically used?

    1 answer
    1. Calcein-AM is a cell-permeant and non-fluorescent compound that is widely used for determining cell viability. In live cells the non-fluorescent calcein-AM is hydrolyzed by intracellular esterases into the strongly green fluorescent anion calcein. The fluorescent calcein is well-retained in the cytoplasm in live cells.

      Helpful?

  7. What is the solubility of Calcein-AM, Product 17783?

    1 answer
    1. Sigma tests the solubility of Product No. 17783 at a concentration of 10 mg/mL in dimethyl sulfoxide (DMSO).

      Helpful?

  8. Do the Scienceware® brand polypropylene products contain any leachable material?

    1 answer
    1. Although Bel-Art does not specifically test their Scienceware® brand polypropylene products, they have no evidence of any problems with leachable material in the polypropylene.

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service