Skip to Content
Merck
All Photos(1)

Documents

17772

Sigma-Aldrich

Retinol

≥95.0% (HPLC), ~2700 U/mg

Synonym(s):

Axerophthol, Vitamin A, Vitamin A alcohol, Vitamin A1, all-trans-3,7-Dimethyl-9-(2,6,6-trimethyl-1-cyclohexen-1-yl)-2,4,6,8-nonatetraen-1-ol

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C20H30O
CAS Number:
Molecular Weight:
286.45
Beilstein:
403040
EC Number:
MDL number:
UNSPSC Code:
12352205
eCl@ss:
34058018
PubChem Substance ID:
NACRES:
NA.79

biological source

synthetic

Assay

≥95.0% (HPLC)

form

(Powder or Crystal with Lumps)

specific activity

~2700 U/mg

color

yellow to dark yellow

mp

61-63 °C (lit.)

shipped in

dry ice

storage temp.

−20°C

SMILES string

CC1(C)C(/C=C/C(C)=C/C=C/C(C)=C/CO)=C(C)CCC1

InChI

1S/C20H30O/c1-16(8-6-9-17(2)13-15-21)11-12-19-18(3)10-7-14-20(19,4)5/h6,8-9,11-13,21H,7,10,14-15H2,1-5H3/b9-6+,12-11+,16-8+,17-13+

InChI key

FPIPGXGPPPQFEQ-OVSJKPMPSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Retinal, retinol and retinoic acid are the aldehyde, alcohol and acid forms of vitamin A. The retinoids exist as many geometric isomers due to the unsaturated bonds in the aliphatic chain. Retinol is biologically active in a wide range of processes.
Retinol or all-trans-retinol is a 20-carbon molecule with cyclohexenyl ring, a side chain with four double bonds (all in trans configuration) and an alcohol end group.

Application

Retinol has been used as a standard in ultra-performance liquid chromatography - tandem mass spectrometer (UPLC-MS/MS) for the vitamins detection and quantification.

Biochem/physiol Actions

Retinol and its derivatives exhibit anti-aging properties. Retinol is used for treating wrinkles and signs of aging. However, due to its photo instability and skin irritation potency, it is hardly used in cosmetic formulations. Retinol is also used as a therapeutic for dermatoses. Its deficiency leads to xerosis and follicular hyperkeratosis.

Unit Definition

1 U corresponds to 1 international U acc. to Ph Eur II, 217 (1983)

Pictograms

Health hazardExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Aquatic Chronic 4 - Eye Irrit. 2 - Repr. 1B - Skin Sens. 1

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 1

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Effects of dietary lead exposure on vitamin levels in great tit nestlings-An experimental manipulation
Ruiz S, et al.
Environmental Pollution (Barking, Essex : 1987), 213(2), 688-697 (2016)
Jacqueline A Jumpsen et al.
Journal of cystic fibrosis : official journal of the European Cystic Fibrosis Society, 5(2), 77-84 (2006-03-02)
The objective of this study was to investigate the effect of docosahexaenoic acid (DHA) supplementation on blood and intestinal DHA levels and lung function in mild/moderately affected adult CF patients with the DeltaF508 genotype. Cystic Fibrosis (CF) patients often present
S P Spencer et al.
Science (New York, N.Y.), 343(6169), 432-437 (2014-01-25)
How the immune system adapts to malnutrition to sustain immunity at barrier surfaces, such as the intestine, remains unclear. Vitamin A deficiency is one of the most common micronutrient deficiencies and is associated with profound defects in adaptive immunity. Here
K R Norum et al.
The American journal of clinical nutrition, 56(4), 735-744 (1992-10-01)
We discuss vitamin A with emphasis on its absorption, transport, cellular uptake, storage, and intracellular metabolism. Dietary retinyl esters are hydrolyzed to retinol in the intestinal lumen before absorption by enterocytes. Carotenoids are absorbed and then partially converted to retinol
Retinoids in the treatment of skin aging: an overview of clinical efficacy and safety
Mukherjee S, et al.
Clinical Interventions in Aging, 1(4), 327-327 (2006)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service