Skip to Content
Merck
All Photos(1)

Key Documents

16859

Sigma-Aldrich

D-α-Hydroxyglutaric acid disodium salt

≥98.0% (GC)

Synonym(s):

(R)-2-Hydroxypentanedioic acid disodium salt, Disodium (R)-2-hydroxyglutarate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C5H6O5Na2
CAS Number:
Molecular Weight:
192.08
Beilstein:
5318041
MDL number:
UNSPSC Code:
12352106
PubChem Substance ID:
NACRES:
NA.25

Quality Level

Assay

≥98.0% (GC)

form

powder or crystals

optical activity

[α]/D 8.5±1.5°, c = 1 in NaOH

impurities

≤6.0% water

storage temp.

2-8°C

SMILES string

[Na].O[C@H](CCC(O)=O)C(O)=O

InChI

1S/C5H8O5.2Na/c6-3(5(9)10)1-2-4(7)8;;/h3,6H,1-2H2,(H,7,8)(H,9,10);;/q;2*+1/p-2/t3-;;/m1../s1

InChI key

DZHFTEDSQFPDPP-HWYNEVGZSA-L

Looking for similar products? Visit Product Comparison Guide

Biochem/physiol Actions

Biomarker for inborn errors of metabolism and cancer

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Seth J Parker et al.
Pharmacology & therapeutics, 152, 54-62 (2015-05-10)
Specific point mutations in isocitrate dehydrogenase 1 and 2 (IDH1 and IDH2) occur in a variety of cancers, including acute myeloid leukemia (AML), low-grade gliomas, and chondrosarcomas. These mutations inactivate wild-type enzymatic activity and convey neomorphic function to produce d-2-hydroxyglutarate
Stefan Nowicki et al.
The FEBS journal, 282(15), 2796-2805 (2015-04-14)
Increased glucose metabolism in cancer cells is a phenomenon that has been known for over 90 years, allowing maximal cell growth through faster ATP production and redistribution of carbons towards nucleotide, protein and fatty acid synthesis. Recently, metabolites that can
Andrew J Worth et al.
Chemical research in toxicology, 28(5), 948-954 (2015-03-25)
The α-ketoglutarate metabolite, 2-hydroxyglutarate (2-HG), has emerged as an important mediator in a subset of cancers and rare inherited inborn errors of metabolism. Because of potential enantiospecific metabolism, chiral analysis is essential for determining the biochemical impacts of altered 2-HG
Stereospecific ketonization of 2-hydroxymuconate by 4-oxalocrotonate tautomerase and 5-(carboxymethyl)-2-hydroxymuconate isomerase.
Whitman, C.P., et al.
Journal of the American Chemical Society, 114, 10104-10110 (1992)
Stereochemical studies on porphyrin a: assignment of the absolute configuration of a model porphyrin by degradation.
Battersby, A.R., et al.
Journal of the Chemical Society. Perkin Transactions 1, 9, 1565-1580 (1986)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service