Skip to Content
Merck
All Photos(1)

Documents

16805

Sigma-Aldrich

Atto 425 NHS ester

BioReagent, suitable for fluorescence, ≥90% (HPLC)

Synonym(s):

Atto 425-N-hydroxysuccinimide ester

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C26H30N2O8
CAS Number:
Molecular Weight:
498.53
MDL number:
UNSPSC Code:
12352108
PubChem Substance ID:
NACRES:
NA.32

product line

BioReagent

Assay

≥90% (HPLC)
≥90% (degree of coupling)

manufacturer/tradename

ATTO-TEC GmbH

solubility

DMF: soluble
DMSO: soluble
H2O: soluble

λ

in ethanol (with 0.1% trifluoroacetic acid)

UV absorption

λ: 422-428 nm Amax

suitability

suitable for fluorescence

storage temp.

−20°C

SMILES string

CCOC(=O)C1=Cc2cc3C(C)CC(C)(C)N(CCCC(=O)ON4C(=O)CCC4=O)c3cc2OC1=O

InChI

1S/C26H30N2O8/c1-5-34-24(32)18-12-16-11-17-15(2)14-26(3,4)27(19(17)13-20(16)35-25(18)33)10-6-7-23(31)36-28-21(29)8-9-22(28)30/h11-13,15H,5-10,14H2,1-4H3

InChI key

PUEQEMDTFPYCDY-UHFFFAOYSA-N

General description

Atto 425 is a member of the commercially available coumarin derivatives.

Application

Atto 425 NHS ester is used in the development of bifunctional fluorescent, inorganic micro-partices (Zeolite L crystals) designed for targeting or imaging mutliple components . Reacts with amino groups or amine modified oliognucleotides to form a stable amine bond. Utilized as an essential component of molecular beacons, which were designed for use in real-time PCR assays to allow for rapid and accurate identification of gram-negative bacteria . When conjugated with benzylguanine, the fluorescence of Atto 425-NHS is quenched but becomes fluorescent upon labeling of the SNAP-tag fusion protein, making this conjugate an ideal candidate for experiments in living cells . Used as one of the donor probes in three-color FRET experiments designed to investigate the kinetics of RNA-protein complex formation .
Atto fluorescent labels are designed for high sensitivity applications, including single molecule detection. Atto labels have rigid structures that do not show any cis-trans-isomerization. Thus these labels display exceptional intensity with minimal spectral shift on conjugation.

Other Notes

Study of stability during one- and two-photon excitation in fluorescence correlation spectroscopy, FCS; employed in triple-color coincidence analysis.

Legal Information

This product is for Research use only. In case of intended commercialization, please contact the IP-holder (ATTO-TEC GmbH, Germany) for licensing.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

P.S. Dittrich, P. Schwille
Applied Physics B: Lasers and Optics, 73, 829-829 (2001)
Handbook of Biophotonics: Vol. 2: Photonics for Health Care (2011)
Frequency domain fluorescence microspectrometry: Application to cellular uptake and drug distribution.
Praus, P., et al.
Spectroscopy: An International Journal, 24, 303-307 (2010)
Nermin Seda Kehr et al.
Nanoscale, 2(4), 601-605 (2010-07-21)
Microcontact printing (mCP) is used to immobilize dyes and peptides asymmetrically, by a "peptide coupling" reaction, on monolayers of zeolite L crystals in the contact area between the stamp and the surface of the monolayer. Chemically patterned surfaces of monolayers
Richard S Agnes et al.
Journal of the American Chemical Society, 132(17), 6075-6080 (2010-04-13)
A fluorescent sensor of protein kinase activity has been developed and used to characterize the compartmentalized location of cAMP-dependent protein kinase activity in mitochondria. The sensor functions via a phosphorylation-induced release of a quencher from a peptide-based substrate, producing a

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service