14531C
EX-CELL® EBx™ PRO-I Serum-Free Media
Animal-component free, without L-glutamine, without phenol red, liquid, suitable for cell culture
Pharma Manufacturing
Synonym(s):
EX-CELL® EBx™ Serum-Free Media
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
UNSPSC Code:
12352207
NACRES:
NA.75
Recommended Products
Quality Level
description
Animal-component-free, low protein
for research or for further manufacturing use
form
liquid
availability
not available in Japan
technique(s)
cell culture | mammalian: suitable
components
L-glutamine: no
phenol red: no
shipped in
wet ice
storage temp.
2-8°C
2-8°C
Looking for similar products? Visit Product Comparison Guide
Legal Information
EBx is a trademark of Vivalis SA
EX-CELL is a registered trademark of Merck KGaA, Darmstadt, Germany
Storage Class Code
12 - Non Combustible Liquids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service