Skip to Content
Merck
All Photos(1)

Key Documents

11625

Sigma-Aldrich

Di-tert-butyl azodicarboxylate

purum, ≥98.0% (GC)

Synonym(s):

Bis(1,1-dimethylethyl)azodicarboxylate, DBAD, Di-tert-butyl azodiformate, NSC 109889

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CH3)3COCON=NCOOC(CH3)3
CAS Number:
Molecular Weight:
230.26
Beilstein:
1911434
EC Number:
MDL number:
UNSPSC Code:
12352108
PubChem Substance ID:
NACRES:
NA.22

grade

purum

Quality Level

Assay

≥98.0% (GC)

form

solid

mp

89-92 °C (lit.)
89-92 °C

functional group

azo

storage temp.

2-8°C

SMILES string

CC(C)(C)OC(=O)\N=N\C(=O)OC(C)(C)C

InChI

1S/C10H18N2O4/c1-9(2,3)15-7(13)11-12-8(14)16-10(4,5)6/h1-6H3/b12-11+

InChI key

QKSQWQOAUQFORH-VAWYXSNFSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Di-tert-butyl azodicarboxylate is a an acid labile reagent for Mitsunobu reactions allowing facile isolation of products and for the electrophilic amination and hydrazination of enolates and lithium alkyls.

Application

Reactant for:
  • Preparation of hexapeptide key fragments via stereoselective selenocyclization/oxidative deselenylation or hydrazination/cyclization reactions
  • Asymmetric Michael addition reactions
  • Preparation of dipeptidyl peptidase IV dependent water-soluble prodrugs via Mitsunobu reaction
  • Synthesis of pyrroloisoquinoline template via stereoselective N-acyliminium-mediated cyclization and enolate amination for synthesis of peptidomimetic compounds
  • Barbier-type propargylation reactions
  • Synthesis of bacterial peptide deformylase (PDF) inhibitor fumimycin
  • Asymmetric amination of glycine Schiff bases

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Masaru Kondo et al.
Angewandte Chemie (International ed. in English), 54(28), 8198-8202 (2015-05-28)
The catalytic enantioselective reaction of diphenylmethylidene-protected α-aminoacetonitriles with imines has been developed. Good yields and diastereo- and enantioselectivities were observed for the reaction of various imines using chiral bis(imidazoline)/Pd catalysts. The reaction of α-aminonitriles with di-tert-butyl azodicarboxylate afforded chiral α,α-diaminonitriles
M. Kiankarimi et al.
Tetrahedron Letters, 40, 4497 -4497 (1999)
J.L. Vicario et al.
Tetrahedron Letters, 40, 7123 -7123 (1999)
D.A. Evans et al.
Journal of the American Chemical Society, 108, 6395-6395 (1986)
C. Gennari et al.
Journal of the American Chemical Society, 108, 6394-6394 (1986)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service