Skip to Content
Merck
All Photos(1)

Key Documents

10385

Sigma-Aldrich

Angiotensin III trifluoroacetate salt hydrate

≥98% (HPCE)

Synonym(s):

Des-Asp-1-Angiotensin II trifluoroacetate salt hydrate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C46H66N12O9 · xC2HF3O2 · yH2O
CAS Number:
Molecular Weight:
931.09 (anhydrous free base basis)
Beilstein:
8385695
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.77
Pricing and availability is not currently available.

Assay

≥98% (HPCE)

solubility

H2O: 1.0 mg/mL, clear, colorless

storage temp.

−20°C

SMILES string

O.OC(=O)C(F)(F)F.CC[C@H](C)[C@H](NC(=O)[C@H](Cc1ccc(O)cc1)NC(=O)[C@@H](NC(=O)[C@@H](N)CCCNC(N)=N)C(C)C)C(=O)N[C@@H](Cc2c[nH]cn2)C(=O)N3CCC[C@H]3C(=O)N[C@@H](Cc4ccccc4)C(O)=O

InChI

1S/C46H66N12O9.C2HF3O2.H2O/c1-5-27(4)38(57-40(61)33(21-29-15-17-31(59)18-16-29)53-42(63)37(26(2)3)56-39(60)32(47)13-9-19-51-46(48)49)43(64)54-34(23-30-24-50-25-52-30)44(65)58-20-10-14-36(58)41(62)55-35(45(66)67)22-28-11-7-6-8-12-28;3-2(4,5)1(6)7;/h6-8,11-12,15-18,24-27,32-38,59H,5,9-10,13-14,19-23,47H2,1-4H3,(H,50,52)(H,53,63)(H,54,64)(H,55,62)(H,56,60)(H,57,61)(H,66,67)(H4,48,49,51);(H,6,7);1H2/t27-,32-,33-,34-,35-,36-,37-,38-;;/m0../s1

InChI key

JEOAJMGVSAMVML-UFPGTABSSA-N

Amino Acid Sequence

Arg-Val-Tyr-Ile-His-Pro-Phe

General description

Angiotensin III (Ang III) is a 7-AA bioactive peptide that is formed from the degradation of the Angiotensin II peptide by aminopeptidase A; both are derivatives of angiotensinogen and are components of the renin-angiotensin system (RAS).

Biochem/physiol Actions

Angiotensin III is an AT1 and AT2 agonist, with higher sensitivity for the AT2 receptor.[1] Ang III has been shown to be a main effector peptide in RAS-controlled vasopresin release[2] and a stimulator for aldosterone release.[3]

Features and Benefits

This compound is featured on the Angiotensin Receptors page of the Handbook of Receptor Classification and Signal Transduction. To browse other handbook pages, click here.

Antibody Adsorbant

Arg-Val-Tyr-Ile-His-Pro-Phe-OH.3CH3COOH.4H2O

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Kenji Oki et al.
Endocrinology, 154(1), 214-221 (2012-12-12)
Aldosterone is synthesized in the zona glomerulosa of the adrenal cortex under primary regulation by the renin-angiotensin system. Angiotensin II (A-II) acts through the angiotensin types 1 and 2 receptors (AT1R and AT2R). A-II is metabolized in different tissues by
Kamila Domińska et al.
Endokrynologia Polska, 60(5), 363-369 (2009-11-04)
The results of many studies have reported that peptides from the angiotensin family are involved in the regulation of cell growth, proliferation, cell migration, apoptosis, inflammation, differentiation, and angiogenesis, which suggests that they might play an important role in carcinogenesis.
Rui Yang et al.
Hypertension research : official journal of the Japanese Society of Hypertension, 34(6), 674-683 (2011-03-18)
Angiotensin (Ang) II, the main effector peptide of the renin-Ang system, increases arterial blood pressure through Ang II type 1A (AT(1a)) receptor-dependent arterial vasoconstriction and by decreasing renal salt and water excretion through extrarenal and intrarenal mechanisms. AT(2) receptors are
Maria del Pilar Carrera et al.
Tumour biology : the journal of the International Society for Oncodevelopmental Biology and Medicine, 31(6), 583-588 (2010-07-24)
Angiotensin II in particular and/or the local renin-angiotensin system in general could have an important role in epithelial tissue growth and modelling; therefore, it is possible that it may be involved in breast cancer. In this sense, previous works of
Sadia Rabbani et al.
Analytical chemistry, 83(10), 3793-3800 (2011-04-06)
Time-of-flight secondary ion mass spectrometry (TOF-SIMS) is an established technique for the characterization of solid sample surfaces. The introduction of polyatomic ion beams, such as C(60), has provided the associated ability to perform molecular depth-profiling and 3D molecular imaging. However

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service