Skip to Content
Merck

08561

(S)-(−)-α -Lipoic acid

≥97% (HPLC)

Synonym(s):

(S)-1,2-Dithiolane-3-pentanoic acid, (S)-6,8-Thioctic acid

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Empirical Formula (Hill Notation):
C8H14O2S2
CAS Number:
Molecular Weight:
206.33
NACRES:
NA.25
PubChem Substance ID:
UNSPSC Code:
12352106
MDL number:
Beilstein/REAXYS Number:
81852
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Quality Segment

assay

≥97% (HPLC)

optical purity

enantiomeric excess: ≥98.0% (HPLC)

storage temp.

−20°C

InChI

1S/C8H14O2S2/c9-8(10)4-2-1-3-7-5-6-11-12-7/h7H,1-6H2,(H,9,10)/t7-/m0/s1

InChI key

AGBQKNBQESQNJD-ZETCQYMHSA-N

Biochem/physiol Actions

Biological antioxidant with prooxidant activities . Method for enantioseparation of lipoic acid in dietary supplements by capillary electrophoresis.

Packaging

Bottomless glass bottle. Contents are inside inserted fused cone.


pictograms

Exclamation mark

signalword

Warning

hcodes

Hazard Classifications

Acute Tox. 4 Oral

Storage Class

11 - Combustible Solids

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library