Skip to Content
Merck
All Photos(1)

Key Documents

S8263

Sigma-Aldrich

Sodium hydroxide solution

5.0 M

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
NaOH
CAS Number:
Molecular Weight:
40.00
MDL number:
UNSPSC Code:
12352316
PubChem Substance ID:
NACRES:
NA.21

grade

Molecular Biology
for molecular biology

Agency

suitable for SM 4500 - NH3

vapor pressure

3 mmHg ( 37 °C)

form

liquid

concentration

5.0 M

pH

14

density

1.327 g/cm3 at 25 °C

storage temp.

room temp

SMILES string

[OH-].[Na+]

InChI

1S/Na.H2O/h;1H2/q+1;/p-1

InChI key

HEMHJVSKTPXQMS-UHFFFAOYSA-M

Looking for similar products? Visit Product Comparison Guide

Application

Sodium hydroxide solution can be used:
  • As a base in the reduction of oximes, imines, and hydrazones using Raney nickel.
  • In the preparation of trans-1,2-cyclohexanediaminetetraacetic acid standard solution applicable in the synthesis of the ferric chelate complex.
  • In the synthesis of radiolabeled quinolinyl analogs for α-synuclein binding assay studies.

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Dam. 1 - Met. Corr. 1 - Skin Corr. 1A

Storage Class Code

8B - Non-combustible corrosive hazardous materials

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Convenient method for reduction of CN double bonds in oximes, imines, and hydrazones using sodium borohydride-Raney Ni system
Yang Y, et al.
Synthetic Communications, 42, 2540-2554 (2012)
Solubility of dimethyldisulfide (DMDS) in aqueous solutions of Fe (III) complexes of trans-1, 2-cyclohexanediaminetetraacetic acid (CDTA) using the static headspace method
Iliuta MC and Larachi F
Fluid Phase Equilibria, 233, 184-189 (2005)
Design, synthesis, and in vitro evaluation of quinolinyl analogues for α-synuclein aggregation
Yue X, et al.
Bioorganic & medicinal chemistry letters, 28, 1011-1019 (2018)
Miki Wada et al.
Nucleic acids research, 42(12), 7851-7866 (2014-06-11)
During termination of translation in eukaryotes, a GTP-binding protein, eRF3, functions within a complex with the tRNA-mimicking protein, eRF1, to decode stop codons. It remains unclear how the tRNA-mimicking protein co-operates with the GTPase and with the functional sites on
Richard J Travers et al.
Blood, 124(22), 3183-3190 (2014-09-10)
Polyphosphate (polyP) is secreted by activated platelets and has been shown to contribute to thrombosis, suggesting that it could be a novel antithrombotic target. Previously reported polyP inhibitors based on polycationic substances, such as polyethylenimine, polyamidoamine dendrimers, and polymyxin B

Related Content

This page is intended to make it easier to find the consumables you need based on the analytical method you’re using. Methods included on this page come from the EPA, Standard Methods and ASTM.

This page is intended to make it easier to find the consumables you need based on the analytical method you’re using. Methods included on this page come from the EPA, Standard Methods and ASTM.

This page is intended to make it easier to find the consumables you need based on the analytical method you’re using. Methods included on this page come from the EPA, Standard Methods and ASTM.

This page is intended to make it easier to find the consumables you need based on the analytical method you’re using. Methods included on this page come from the EPA, Standard Methods and ASTM.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service