Skip to Content
Merck
All Photos(2)

Key Documents

M46837

Sigma-Aldrich

Methyl formate

reagent grade, 97%

Synonym(s):

Formic acid methyl ester

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
HCO2CH3
CAS Number:
Molecular Weight:
60.05
Beilstein:
1734623
EC Number:
MDL number:
UNSPSC Code:
12352108
PubChem Substance ID:
NACRES:
NA.21
grade:
reagent grade
Assay:
97%
bp:
32-34 °C (lit.)
vapor pressure:
32.81 psi ( 55 °C)
9.21 psi ( 20 °C)
Pricing and availability is not currently available.

grade

reagent grade

Quality Level

vapor density

2.1 (vs air)

vapor pressure

32.81 psi ( 55 °C)
9.21 psi ( 20 °C)

Assay

97%

form

liquid

autoignition temp.

842 °F

expl. lim.

23 %

impurities

≤3% methyl alcohol

refractive index

n20/D 1.343 (lit.)

pH

4.0-5.0 (20 °C, 200 g/L)

bp

32-34 °C (lit.)

mp

−100 °C (lit.)

density

0.974 g/mL at 20 °C (lit.)

SMILES string

[H]C(=O)OC

InChI

1S/C2H4O2/c1-4-2-3/h2H,1H3

InChI key

TZIHFWKZFHZASV-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Methyl formate may be used to prepare formamides via amine formaylation.[1]

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Eye Irrit. 2 - Flam. Liq. 1 - STOT SE 1 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

-2.2 °F - closed cup

Flash Point(C)

-19 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Marine S Da Silva et al.
Prostaglandins, leukotrienes, and essential fatty acids, 126, 64-71 (2017-10-17)
This study aimed to determine whether dairy macronutrients alter markers of inflammation and oxidative stress in endothelial cells. Human endothelial cells (HUVEC) were treated with ruminant trans fatty acids (rTFA), either trans-vaccenic acid (tVA) or trans-palmitoleic acid (tPA), whey protein
Methyl formate as a new building block in C1 chemistry.
Lee JS, et al.
Applied Catalysis, 57(1), 1-30 (1990)
Rotational isomerism in methyl formate and methyl acetate; a low-temperature matrix infrared study using thermal molecular beams.
Blom CE and Gunthard HH.
Chemical Physics Letters, 84(2), 267-271 (1981)
Catalytic reaction of methyl formate with amines to formamides.
Deutsch J, et al.
Tetrahedron, 65(50), 10365-10369 (2009)
Jean-François Bilodeau et al.
Prostaglandins, leukotrienes, and essential fatty acids, 164, 102215-102215 (2020-12-05)
Cancer has been associated with increased oxidative stress and deregulation of bioactive oxylipins derived from long-chain polyunsaturated fatty acids (LC-PUFA) like arachidonic acid (AA). There is a debate whether ω-3 LC-PUFA could promote or prevent prostate tumor growth through immune

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service