Skip to Content
Merck
All Photos(1)

Key Documents

71948

Sigma-Aldrich

Sodium selenate

purum p.a., ≥98.0% (T)

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
Na2SeO4
CAS Number:
Molecular Weight:
188.94
EC Number:
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:
NACRES:
NA.21
form:
powder or crystals
Pricing and availability is not currently available.

grade

purum p.a.

Quality Level

Assay

≥98.0% (T)

form

powder or crystals

pH

5.5-7.5 (25 °C, 18.9 g/L)

anion traces

chloride (Cl-): ≤1000 mg/kg

cation traces

Ca: ≤200 mg/kg
Cd: ≤50 mg/kg
Co: ≤50 mg/kg
Cu: ≤50 mg/kg
Fe: ≤50 mg/kg
K: ≤1000 mg/kg
Ni: ≤50 mg/kg
Pb: ≤50 mg/kg
Zn: ≤50 mg/kg

SMILES string

[Na+].[Na+].[O-][Se]([O-])(=O)=O

InChI

1S/2Na.H2O4Se/c;;1-5(2,3)4/h;;(H2,1,2,3,4)/q2*+1;/p-2

InChI key

MHQOTKLEMKRJIR-UHFFFAOYSA-L

Looking for similar products? Visit Product Comparison Guide

Related Categories

General description

Sodium selenite (Na2SeO4) is a selenium (VI) salt.[1]

Application

Sodium selenite (Na2SeO4) may be employed as a catalyst during the green protocol for synthesis of tetrahydrobenzo[b]pyran derivatives.[2]

Signal Word

Danger

Hazard Classifications

Acute Tox. 2 Inhalation - Acute Tox. 2 Oral - Aquatic Acute 1 - Aquatic Chronic 1 - Skin Irrit. 2 - STOT RE 2

Storage Class Code

6.1B - Non-combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Sodium selenate catalyzed simple and efficient synthesis of tetrahydro benzo [b] pyran derivatives.
Hekmatshoar R, et al.
Catalysis Communications, 9(2), 307-310 (2008)
Biological and chemical investigation of Allium cepa L. response to selenium inorganic compounds.
Michalska-Kacymirow M, et al.
Analytical and Bioanalytical Chemistry, 406(15), 3717-3722 (2015)
Maria Łabanowska et al.
Journal of plant physiology, 169(3), 275-284 (2011-12-14)
Biochemical analyses of antioxidant content were compared with measurements of fluorescence and electron paramagnetic resonance (EPR) to examine the alteration of radicals in wheat seedlings exposed to 2 days of selenium stress. Two genotypes of Polish and one of Finnish
M Djanaguiraman et al.
Plant physiology and biochemistry : PPB, 48(12), 999-1007 (2010-10-19)
Oxidative stress is commonly induced when plants are grown under high temperature (HT) stress conditions. Selenium often acts as an antioxidant in plants; however, its role under HT-induced oxidative stress is not definite. We hypothesize that selenium application can partly
Alexandra Holmstrom et al.
The Journal of nutritional biochemistry, 23(9), 1086-1091 (2011-12-06)
The inhibitory effect of oral methylseleninic acid or methylselenocysteine administration on cancer cell xenograft development in nude mice is well characterized; however, less is known about the efficacy of selenate and age on selenium chemoprevention. In this study, we tested

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service