71650
Sodium phosphate dibasic dodecahydrate
puriss. p.a., crystallized, ≥99.0% (T)
Synonym(s):
sec-Sodium phosphate, Disodium hydrogen phosphate dodecahydrate, Disodium phosphate, di-Sodium hydrogen phosphate dodecahydrate
About This Item
Recommended Products
grade
puriss. p.a.
Assay
≥99.0% (T)
form
crystals
quality
crystallized
pH
9.0-9.3 (25 °C, 50 mg/mL in H2O)
solubility
water: soluble 35.8 g/L at 20 °C
anion traces
chloride (Cl-): ≤5 mg/kg
fluoride (F-): ≤5 mg/kg
sulfate (SO42-): ≤50 mg/kg
cation traces
As: ≤0.5 mg/kg
Ca: ≤10 mg/kg
Cd: ≤5 mg/kg
Co: ≤5 mg/kg
Cr: ≤5 mg/kg
Cu: ≤5 mg/kg
Fe: ≤5 mg/kg
K: ≤50 mg/kg
Mg: ≤5 mg/kg
Mn: ≤5 mg/kg
Ni: ≤5 mg/kg
Pb: ≤5 mg/kg
Zn: ≤5 mg/kg
SMILES string
O.O.O.O.O.O.O.O.O.O.O.O.[Na+].[Na+].OP([O-])([O-])=O
InChI
1S/2Na.H3O4P.12H2O/c;;1-5(2,3)4;;;;;;;;;;;;/h;;(H3,1,2,3,4);12*1H2/q2*+1;;;;;;;;;;;;;/p-2
InChI key
DGLRDKLJZLEJCY-UHFFFAOYSA-L
Looking for similar products? Visit Product Comparison Guide
General description
Application
Storage Class Code
13 - Non Combustible Solids
WGK
WGK 1
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service