Skip to Content
Merck
All Photos(1)

Key Documents

537616

Sigma-Aldrich

Diethylene glycol monoethyl ether

ReagentPlus®, 99%

Synonym(s):

2-(2-Ethoxyethoxy)ethanol, CARBITOL, Diethylene glycol ethyl ether, Ethyldiglycol

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C2H5OCH2CH2OCH2CH2OH
CAS Number:
Molecular Weight:
134.17
Beilstein:
1736441
EC Number:
MDL number:
UNSPSC Code:
12352112
PubChem Substance ID:
NACRES:
NA.21
Assay:
99%
bp:
202 °C (lit.)
vapor pressure:
0.12 mmHg ( 20 °C)
Pricing and availability is not currently available.

vapor density

4.63 (vs air)

Quality Level

vapor pressure

0.12 mmHg ( 20 °C)

product line

ReagentPlus®

Assay

99%

form

liquid

expl. lim.

1.2 %, 135 °F
23.5 %, 182 °F

refractive index

n20/D 1.427 (lit.)

bp

202 °C (lit.)

mp

-80 °C

solubility

water: soluble

density

0.999 g/mL at 25 °C (lit.)

SMILES string

CCOCCOCCO

InChI

1S/C6H14O3/c1-2-8-5-6-9-4-3-7/h7H,2-6H2,1H3

InChI key

XXJWXESWEXIICW-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Diethylene glycol monoethyl ether (DEGMEE) is an organic solvent commonly used in the cosmetic industry.[1] Its toxicity has been assessed.[2] Various thermodynamic and optical parameters of an aqueous solution of DEGMEE have been determined.[3] Its quantification along with diethylene glycol (DEG) and coumarin in food samples by gas chromatography-flame ionization technique has been reported.[4]

Application


  • A cautionary note on the effects of laboratory air contaminants on ambient ionization mass spectrometry measurements.: Highlights the influence of diethylene glycol monoethyl ether as a common laboratory contaminant that can affect the accuracy of mass spectrometry analyses, underscoring the importance of controlling environmental variables in high-precision analytical settings (Kumbhani SR et al., 2017).

Legal Information

ReagentPlus is a registered trademark of Merck KGaA, Darmstadt, Germany
CARBITOL is a trademark of The Dow Chemical Company or an affiliated company of Dow

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

204.8 °F - closed cup

Flash Point(C)

96 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Thermodynamic and optical studies of some ethylene glycol ethers in aqueous solutions at T = 298.15 K.
Dhondge SS, et al.
The Journal of Chemical Thermodynamics, 41(5), 577-585 (2009)
Simultaneous determination of diethylene glycol, diethylene glycol monoethyl ether, coumarin and caffeine in food items by gas chromatography.
Rahim AA, et al.
Food Chemistry, 126(3), 1412-1416 (2011)
Reproductive toxicology. Diethylene glycol monoethyl ether.
Environmental health perspectives, 105 Suppl 1, 209-210 (1997-02-01)
Hyun-Jong Cho et al.
International journal of pharmaceutics, 423(2), 153-160 (2012-01-03)
To achieve rapid onset of action and improved bioavailability of udenafil, a microemulsion system was developed for its intranasal delivery. Phase behavior, particle size, transmission electron microscope (TEM) images, and the drug solubilization capacity of the microemulsion were investigated. A
Simona Mura et al.
Journal of drug targeting, 19(3), 189-196 (2010-05-08)
The aim of this work was to evaluate the ability of Transcutol (Trc) to produce elastic vesicles with soy lecithin (SL) and study the influence of the obtained vesicles on in vitro (trans)dermal delivery of minoxidil. To this purpose, so-called

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service