Skip to Content
Merck
All Photos(4)

Key Documents

393703

Sigma-Aldrich

Chromium(III) oxide

powder, ≥98%

Synonym(s):

Chromia

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
Cr2O3
CAS Number:
Molecular Weight:
151.99
EC Number:
MDL number:
UNSPSC Code:
12352303
PubChem Substance ID:
NACRES:
NA.55

Quality Level

Assay

≥98%

form

powder

SMILES string

O=[Cr]O[Cr]=O

InChI

1S/2Cr.3O

InChI key

QDOXWKRWXJOMAK-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Chromium (III) oxide is a chromium complex in which the chromium ion is in +3 oxidation state. The synthesis of its sub-micron powder has been reported.† The IR and Raman spectra of chromium (III) oxide have been studied.† The impact of adding lysozyme on the stability of chromium (III) oxide (Cr2O3) suspension has been evaluated.†

Application

Chromium (III) oxide may be used in the following processes:
  • To modify carbon paste electrodes for optimizing its ability to detect nitric oxide.
  • Preparation of Ni+Cr2O3 composite coatings.
  • Formation of the ternary oxide, CrVMoO7 by solid-state reaction which may be employed in selective oxidation reactions.
Used in a solid-state reaction to form the new ternary oxide, CrVMoO7, can be used in selective oxidation.

Storage Class Code

11 - Combustible Solids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

The Hydrogen Evolution Reaction on Ni Electrode Material Modified with Molybdenum (IV) Oxide and Chromium (III) Oxide Powders.
Popczyk M and Losiewicz B.
Solid State Phenomena, 228 (2015)
Amperometric Nitric Oxide Sensor Based on Carbon Paste Electrode Modified with Chromium (III) Oxide.
Berisha LS, et al.
Sensors & Transducers Journal, 184(1), 1726-5479 (2015)
Crystal Structure and Raman Spectroscopy of FeVMoO7 and CrVMoO7 with Mo=O Double Bonds.
Wang, X. et al.
Inorganic Chemistry, 37(13), 3252-3252 (1998)
Jordi Cirera et al.
The Journal of chemical physics, 137(5), 054704-054704 (2012-08-17)
The effects of electronic polarization on the adsorption of water in the MIL-53(Cr) metal-organic framework are investigated using molecular dynamics simulations. For this purpose a fully polarizable force field for MIL-53(Cr) was developed which is compatible with the ab initio-based
Defa Wang et al.
Nano letters, 11(6), 2353-2357 (2011-05-17)
We report on the achievement of wafer-level photocatalytic overall water splitting on GaN nanowires grown by molecular beam epitaxy with the incorporation of Rh/Cr(2)O(3) core-shell nanostructures acting as cocatalysts, through which H(2) evolution is promoted by the noble metal core

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service