Skip to Content
Merck
All Photos(3)

Key Documents

320021

Sigma-Aldrich

3-Methyl-1-butanol

ACS reagent, ≥98.5%

Synonym(s):

Isoamyl alcohol, Isopentyl alcohol

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CH3)2CHCH2CH2OH
CAS Number:
Molecular Weight:
88.15
Beilstein:
1718835
EC Number:
MDL number:
UNSPSC Code:
12352001
PubChem Substance ID:
NACRES:
NA.21
grade:
ACS reagent
bp:
130 °C (lit.)
vapor pressure:
2 mmHg ( 20 °C)

grade

ACS reagent

vapor density

3 (vs air)

vapor pressure

2 mmHg ( 20 °C)

Assay

≥98.5%

form

liquid

autoignition temp.

644 °F

expl. lim.

1.2-9 %, 100 °F

dilution

(for analytical testing)

impurities

≤0.002 meq/g Titr. acid
≤0.1% carbonyl (as HCHO)
≤0.2% acids and esters (as amyl acetate)
≤0.5% water

evapn. residue

≤0.003%

refractive index

n20/D 1.406 (lit.)

pH

5.6 (20 °C, 25 g/L)

bp

130 °C (lit.)

mp

−117 °C (lit.)

density

0.809 g/mL at 25 °C (lit.)

SMILES string

CC(C)CCO

InChI

1S/C5H12O/c1-5(2)3-4-6/h5-6H,3-4H2,1-2H3

InChI key

PHTQWCKDNZKARW-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

3-methyl-1-butanol is a clear, colorless alcohol. It is one of several isomers of amyl alcohol. It is commonly used as a solvent in various applications, such as inks, coatings, and pharmaceutical preparations.

Application

3-Methyl-1-butanol is used as a:
  • Dissolution of a wide range of organic compounds and is often employed for extraction, dilution, and preparation of samples for analysis.
  • Starting material or reagent in organic synthesis. It can be incorporated into various reactions to introduce the isoamyl group or alcohol functionality into target molecules.

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Inhalation - Eye Dam. 1 - Flam. Liq. 3 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

3 - Flammable liquids

WGK

WGK 1

Flash Point(F)

110.3 °F - closed cup

Flash Point(C)

43.5 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Journal of AOAC International
Kim HM, et al.
Journal of AOAC (Association of Official Analytical Chemists) International, 100(5), 1492-1499 (2017)
Pentanol isomer synthesis in engineered microorganisms
Cann AF,et al.
Applied Microbiology and Biotechnology, 85, 893-899 (2010)
Mary J Dunlop et al.
Molecular systems biology, 7, 487-487 (2011-05-11)
Many compounds being considered as candidates for advanced biofuels are toxic to microorganisms. This introduces an undesirable trade-off when engineering metabolic pathways for biofuel production because the engineered microbes must balance production against survival. Cellular export systems, such as efflux
Niels O Verhulst et al.
Malaria journal, 10, 28-28 (2011-02-10)
Anopheles gambiae sensu stricto is considered to be highly anthropophilic and volatiles of human origin provide essential cues during its host-seeking behaviour. A synthetic blend of three human-derived volatiles, ammonia, lactic acid and tetradecanoic acid, attracts A. gambiae. In addition
Jane Melville et al.
PloS one, 9(7), e101847-e101847 (2014-07-31)
Molecular phylogenetics is increasingly highlighting the prevalence of cryptic species, where morphologically similar organisms have long independent evolutionary histories. When such cryptic species are known to be declining in numbers and are at risk of extinction due to a range

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service