Skip to Content
Merck
All Photos(2)

Key Documents

310050

Sigma-Aldrich

Iron(III) oxide

powder, <5 μm, ≥96%

Synonym(s):

Ferric oxide

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
Fe2O3
CAS Number:
Molecular Weight:
159.69
EC Number:
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:
NACRES:
NA.21
grade:
for analytical purposes
form:
powder

grade

for analytical purposes

Assay

≥96%

form

powder

particle size

<5 μm

SMILES string

O=[Fe]O[Fe]=O

InChI

1S/2Fe.3O

InChI key

JEIPFZHSYJVQDO-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Related Categories

General description

Iron (III) oxide (Fe2O3) is an inorganic compound. It is one of the three main types of iron oxide. The other two are the rare iron (II) oxide (FeO) and the naturally occurring iron (II, III) oxide (Fe3O4), found in the mineral magnetite. In the steel industry Fe2O3 is the primary source of iron. In addition to being ferromagnetic, Fe2O3 is easily attacked by acids.

Application

Iron (III) oxide is used:
  • As a catalyst in the reduction of nitroarenes.
  • As an oxidizing agent in the synthesis of [11C] Phosgene ([11C] COCl2).

Storage Class Code

13 - Non Combustible Solids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 2

1 of 2

Single phase, electrically insulating, multiferroic La-substituted BiFeO3 prepared by mechanosynthesis.
Perejon A, et al.
Journal of Material Chemistry C, 2(39), 8398-8411 (2014)
Electrical properties of stoichiometric BiFeO3 prepared by mechanosynthesis with either conventional or spark plasma sintering.
Perejon A, et al.
Journal of the American Ceramic Society. American Ceramic Society, 96(4), 1220-1227 (2013)
Electrochemistry and photoelectrochemistry of iron (III) oxide.
Dare-Edwards MP, et al.
J. Chem. Soc., Faraday, 79(9), 2027-2041 (1983)
Light-induced reduction of natural iron (III) oxide and its relevance to phytoplankton.
Finden DAS, et al.
Nature, 309, 783-784 (1984)
William B Ingler et al.
Journal of the American Chemical Society, 126(33), 10238-10239 (2004-08-19)
Stable zinc-doped iron(III) oxide thin films that exhibit p-type behavior were synthesized by spray pyrolytic deposition (SPD) on conducting indium-doped tin oxide-coated glass substrate. The highest photocurrent density of 1.1 mA/cm2 was observed at an illumination intensity of 40 mW/cm2

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service