Skip to Content
Merck
All Photos(3)

Key Documents

267856

Sigma-Aldrich

Molybdenum(VI) oxide

ACS reagent, ≥99.5%

Synonym(s):

Molybdenum trioxide

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
MoO3
CAS Number:
Molecular Weight:
143.94
EC Number:
MDL number:
UNSPSC Code:
12352303
PubChem Substance ID:
NACRES:
NA.21
Assay:
≥99.5%
grade:
ACS reagent
form:
powder
Pricing and availability is not currently available.

grade

ACS reagent

Quality Level

Assay

≥99.5%

form

powder

reaction suitability

reagent type: catalyst
core: molybdenum

impurities

≤0.01% insol. dil. NH4OH

mp

795 °C (lit.)

anion traces

AsO43-, PO43- and SiO2: ≤0.001%
chloride (Cl-): ≤0.002%
nitrate (NO3-): passes test
phosphate (PO43-): ≤5 ppm
sulfate (SO42-): ≤0.02%

cation traces

NH4+: ≤0.002%
heavy metals (as Pb): ≤0.005%

SMILES string

O=[Mo](=O)=O

InChI

1S/Mo.3O

InChI key

JKQOBWVOAYFWKG-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Precursor to LAMOX fast ion conductors and superconductors.
Used in the solid state synthesis of a remarkable ternary, reduced molybdenum oxide, Pr4Mo9O18, whose structure contains previously unknown Mo7, Mo13 and Mo19 clusters. The new cluster product is a small band gap semiconductor.

Pictograms

Health hazardExclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Carc. 2 - Eye Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Eagleson M.
Concise Encyclopedia Chemistry, 509-509 (1994)
Eagleson M.
Concise Encyclopedia Chemistry, 224-224 (1994)
Kinetics and mechanism of molybdenum (VI) oxide reduction.
Sloczynski J.
Journal of Solid State Chemistry, 118(1), 84-92 (1995)
High-activity MgO-supported CoMo hydrodesulfurization catalysts prepared by non-aqueous impregnation.
Kaluza L, et al.
Applied Catalysis. B, Environmental, 162, 430-436 (2015)
Temperature-programed reduction of molybdenum (VI) oxide and molybdenum (IV) oxide.
Arnoldy P, et al.
The Journal of Physical Chemistry, 89(21), 4517-4526 (1985)

Questions

  1. What is the phase of this material as determined by XRD analysis?

    1 answer
    1. The product is in the orthorhombic phase.

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service