Skip to Content
Merck
All Photos(1)

Key Documents

248630

Sigma-Aldrich

Hydrogen bromide solution

33 wt. % in acetic acid

Synonym(s):

HBr

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
HBr
Molecular Weight:
80.91
MDL number:
UNSPSC Code:
12352106
PubChem Substance ID:
NACRES:
NA.21

vapor density

2.8 (vs air)

Quality Level

vapor pressure

11.4 mmHg ( 20 °C)

form

liquid

concentration

30-32% HBr
33 wt. % in acetic acid

density

1.354 g/mL at 25 °C

SMILES string

Br

InChI

1S/BrH/h1H

InChI key

CPELXLSAUQHCOX-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Physical properties

May darken upon storage.

Pictograms

CorrosionExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Dam. 1 - Met. Corr. 1 - Skin Corr. 1B - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

The use of hydrogen bromide in acetic acid for the removal of carbobenzoxy groups and benzyl esters of peptide derivatives1.
Ben-Ishai DOV.
The Journal of Organic Chemistry, 19(1), 62-66 (1954)
Ahmed Hasbi et al.
FASEB journal : official publication of the Federation of American Societies for Experimental Biology, 28(11), 4806-4820 (2014-07-27)
Although the dopamine D1-D2 receptor heteromer has emerging physiological relevance and a postulated role in different neuropsychiatric disorders, such as drug addiction, depression, and schizophrenia, there is a need for pharmacological tools that selectively target such receptor complexes in order
Anna Partyka et al.
Bioorganic & medicinal chemistry, 23(1), 212-221 (2014-12-02)
Continuing our earlier study in a group of purine-2,6-dione derivatives of long chain arylpiperazines (LCAPs), a series of 8-unsubstituted 7-phenylpiperazin-4-yl-alkyl (4-14) and 7-tetrahydroisoquinolinyl-alkyl (15-17) analogues were synthesized and their serotonin 5-HT1A, 5-HT2A, 5-HT6, 5-HT7 and dopamine D2 receptor affinities were
Yaohui Tang et al.
Journal of cerebral blood flow and metabolism : official journal of the International Society of Cerebral Blood Flow and Metabolism, 34(7), 1138-1147 (2014-04-10)
Neural stem cells (NSCs) show therapeutic potential for ischemia in young-adult animals. However, the effect of aging on NSC therapy is largely unknown. In this work, NSCs were transplanted into aged (24-month-old) and young-adult (3-month-old) rats at 1 day after
Yuri Choi et al.
ACS nano, 8(11), 11377-11385 (2014-10-18)
Hybrid nanoparticles composed of multiple components can offer unique opportunities for understanding the nanoscale mechanism and advanced material applications. Here, we report the synthesis of heterodimeric silver-carbon dot nanoparticles (Ag-CD NPs) where the Ag NP is grown on the surface

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service