Skip to Content
Merck
All Photos(3)

Key Documents

243426

Sigma-Aldrich

Strontium nitrate

ACS reagent, ≥99.0%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
Sr(NO3)2
CAS Number:
Molecular Weight:
211.63
EC Number:
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:
NACRES:
NB.24
Assay:
≥99.0%
grade:
ACS reagent
form:
crystals or chunks

grade

ACS reagent

Quality Level

Assay

≥99.0%

form

crystals or chunks

impurities

≤0.01% insolubles

loss

≤0.1% loss on drying

pH

5.0-7.0 (25 °C, 5%)

mp

570 °C (lit.)

anion traces

chloride (Cl-): ≤0.002%
sulfate (SO42-): ≤0.005%

cation traces

Ba: ≤0.05%
Ca: ≤0.05%
Fe: ≤5 ppm
Mg: ≤0.10%
Na: ≤0.10%
heavy metals: ≤5 ppm (by ICP-OES)

SMILES string

[Sr++].[O-][N+]([O-])=O.[O-][N+]([O-])=O

InChI

1S/2NO3.Sr/c2*2-1(3)4;/q2*-1;+2

InChI key

DHEQXMRUPNDRPG-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Crystal structure analysis of strontium nitrate by single crystal neutron diffraction method suggest that it belongs to the cubic crystal system and space group Pa3.

Application


  • Synthesis and Antimicrobial Analysis of High Surface Area Strontium-Substituted Calcium Phosphate Nanostructures for Bone Regeneration: The study explores the potential of strontium nitrate in bone regeneration through the synthesis of high surface area strontium-substituted calcium phosphate nanostructures with antimicrobial properties (Anwar et al., 2023).
  • Electrochemically Assisted Deposition of Strontium Modified Magnesium Phosphate on Titanium Surfaces: The article describes the use of strontium nitrate in creating coatings on titanium surfaces for improved biomedical applications, enhancing material properties through electrochemical deposition techniques (Meininger et al., 2016).

  • Production of Y-86 and Other Radiometals for Research Purposes Using a Solution Target System: This study involves the use of strontium nitrate in the production of radiometals, including Y-86, for various research and diagnostic purposes, employing a solution target system for effective synthesis (Oehlke et al., 2015).

  • Template-based Fabrication of SrTiO3 and BaTiO3 Nanotubes: The research discusses the utility of strontium nitrate in the template-based fabrication of nanotubes, specifically SrTiO3 and BaTiO3, which are critical in enhancing material properties for various industrial and technological applications (Chen et al., 2009).

Pictograms

Flame over circleCorrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Dam. 1 - Ox. Sol. 1

Storage Class Code

5.1A - Strongly oxidizing hazardous materials

WGK

WGK 1


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Ian S Metcalfe et al.
Nature chemistry, 11(7), 638-643 (2019-05-28)
All real processes, be they chemical, mechanical or electrical, are thermodynamically irreversible and therefore suffer from thermodynamic losses. Here, we report the design and operation of a chemical reactor capable of approaching thermodynamically reversible operation. The reactor was employed for
Structure refinement of strontium nitrate, Sr(NO3)2, and barium nitrate, Ba(NO3)2.
Nowotny H and Heger G.
Acta Crystallographica Section C, Crystal Structure Communications, 39(8), 952-956 (1983)
G S Hahn
Dermatologic surgery : official publication for American Society for Dermatologic Surgery [et al.], 25(9), 689-694 (1999-09-22)
Irritant contact dermatitis occurs following topical exposure to many chemicals found in cosmetics, personal care products, drugs, and during occupational exposure. Signs and symptoms may include sensory irritation (sting, burn, and/or itch), erythema, edema, and vesiculation. In an attempt to
Porous YSZ impregnated with La0.4Sr0.5Ba0.1TiO3 as a possible composite anode for SOFCs fueled with sour feeds.
Vincent AL, et al.
Journal of Power Sources, 215, 301-306 (2012)
H Zhai et al.
Contact dermatitis, 42(2), 98-100 (2000-03-07)
Skin care products are complex formulations that may cause sensory irritation symptoms, characterized by stinging, burning, and itching. Substances capable of counteracting sensory irritation are of great practical interest. Strontium salts have been demonstrated to inhibit sensory irritation and inflammation

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service