Skip to Content
Merck
All Photos(1)

Key Documents

221090

Sigma-Aldrich

Mercury(II) iodide

ACS reagent, ≥99.0%

Synonym(s):

Mercuric iodide red

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
HgI2
CAS Number:
Molecular Weight:
454.40
EC Number:
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:
NACRES:
NA.21
grade:
ACS reagent
form:
powder
solubility:
potassium iodide solution: passes test

grade

ACS reagent

Quality Level

Assay

≥99.0%

form

powder

reaction suitability

reagent type: catalyst
core: mercury

impurities

≤0.05% soluble Hg salts

bp

354 °C (lit.)

mp

259 °C (lit.)

solubility

potassium iodide solution: passes test

cation traces

Hg+: ≤0.1%

SMILES string

I[Hg]I

InChI

1S/Hg.2HI/h;2*1H/q+2;;/p-2

InChI key

YFDLHELOZYVNJE-UHFFFAOYSA-L

Looking for similar products? Visit Product Comparison Guide

General description

Red mercuric oxide possesses the ability to detect high energy radiation (X- and γ-rays) and is used in nuclear radiation detectors.

Application

Mercury(II) iodide may be used in the preparation of the following:
  • mercury(II) acetate
  • di­iodo­bis(1-methyl-1,3-imidazolium-2-thiol­ato-S)­mercury(II)
  • [Hg2(L)(I)4] and [(L′)Hg(μ-I)2HgI2]n (L = N, N′-bis(phenyl(pyridin-2-yl)methylene)propane-1,2-diamine, L′ = N-(phenyl(pyridin-2-yl)methylene)propane-1,2-diamine)

Signal Word

Danger

Hazard Classifications

Acute Tox. 1 Dermal - Acute Tox. 2 Inhalation - Acute Tox. 2 Oral - Aquatic Acute 1 - Aquatic Chronic 1 - STOT RE 2

Storage Class Code

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Preliminary results on mercuric iodide nuclear radiation detectors.
Ponpon JP, et al.
Nuclear Instruments and Methods in Physics Research, 119, 197-198 (1974)
Mercuric iodide for room temperature radiation detectors. Synthesis, purification, crystal growth and defect formation.
Piechotka M.
Materials Science & Engineering. R, Reports : A Review Journal, 18(1), 1-98 (1997)
Syntheses, Characterization, and Crystal Structures of a Dinuclear Complex and Coordination Polymer of Mercury (II) with Schiff Base Ligands containing N3 and N4 Donors.
Abedi M, et al.
Zeitschrift fur Anorganische und Allgemeine Chemie, 640(11), 2193-2197 (2014)
Diiodobis (1-methyl-1, 3-imidazolium-2-thiolato-S) mercury (II).
Pavlovic G, et al.
Acta Crystallographica Section C, Crystal Structure Communications, 56(7), 801-803 (2000)
Eagleson M.
Concise Encyclopedia Chemistry, 637-637 (1994)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service