Skip to Content
Merck
All Photos(1)

Key Documents

20920

Sigma-Aldrich

Cadmium sulfate 8/3-hydrate

puriss. p.a., ACS reagent, ≥99.0% (calc. based on CdSO4 · 8/3 H2O, KT)

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CdSO4 · 8/3H2O
CAS Number:
Molecular Weight:
256.57
EC Number:
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:
NACRES:
NB.24
Assay:
≥99.0% (calc. based on CdSO4 · 8/3 H2O, KT)
form:
powder or crystals

grade

ACS reagent
puriss. p.a.

Quality Level

Assay

≥99.0% (calc. based on CdSO4 · 8/3 H2O, KT)

form

powder or crystals

impurities

≤0.005% insoluble matter in H2O

anion traces

chloride (Cl-): ≤10 mg/kg
nitrate and nitrite (as NO3-): ≤30 mg/kg

cation traces

As: ≤2 mg/kg
Ca: ≤50 mg/kg
Co: ≤5 mg/kg
Cr: ≤5 mg/kg
Cu: ≤5 mg/kg
Fe: ≤5 mg/kg
K: ≤50 mg/kg
Mg: ≤5 mg/kg
Mn: ≤5 mg/kg
Na: ≤50 mg/kg
Ni: ≤10 mg/kg
Pb: ≤20 mg/kg
Zn: ≤50 mg/kg

SMILES string

[Cd++].[Cd++].[Cd++].[H]O[H].[H]O[H].[H]O[H].[H]O[H].[H]O[H].[H]O[H].[H]O[H].[H]O[H].[O-]S([O-])(=O)=O.[O-]S([O-])(=O)=O.[O-]S([O-])(=O)=O

InChI

1S/3Cd.3H2O4S.8H2O/c;;;3*1-5(2,3)4;;;;;;;;/h;;;3*(H2,1,2,3,4);8*1H2/q3*+2;;;;;;;;;;;/p-6

InChI key

AGHQMAQBLOTWPQ-UHFFFAOYSA-H

Looking for similar products? Visit Product Comparison Guide

Related Categories

Signal Word

Danger

Hazard Classifications

Acute Tox. 2 Inhalation - Acute Tox. 3 Oral - Aquatic Acute 1 - Aquatic Chronic 1 - Carc. 1B - Muta. 1B - Repr. 1B - STOT RE 1

Target Organs

Bone,Kidney,Respiratory system

Storage Class Code

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

P Voua Otomo et al.
Ecotoxicology (London, England), 19(2), 362-368 (2009-09-23)
Since life-cycle studies showed that the earthworm species Eisenia fetida can develop increased tolerance after long-term exposure to a sub-lethal concentration of Cd in the laboratory, we assessed both the cytotoxicity and genotoxicity of Cd in a long-term Cd-exposed population.
Ulrico J López-Chuken et al.
Journal of environmental sciences (China), 22(7), 1080-1085 (2010-12-24)
A controlled hydroponic experiment was undertaken to investigate Cd uptake in relation to the activity of Cd species in solution other than the free ion (Cd2+) by maintaining a constant Cd2+ activity under variable SO42- and Cl- concentrations exposed to
Catriona O'Sullivan et al.
Chemical communications (Cambridge, England), 47(9), 2655-2657 (2011-01-12)
A multi component assembly consisting of the redox protein cytochrome c (cyt c) immobilised onto vertically aligned gold tipped semiconductor nanorods is described. Cyt c was successfully immobilised using a thiol linker. A faradaic response demonstrated that the protein is
Laura Saa et al.
Chemistry (Weinheim an der Bergstrasse, Germany), 16(21), 6187-6192 (2010-05-01)
We have developed an analytical assay to detect the enzymatic activity of acetylcholine esterase and alkaline phosphatase based on the generation of quantum dots by enzymatic products. Acetylcholine esterase converts acetylthiocholine into thiocholine. The latter enhances the rate of decomposition
Nina V Kuchkina et al.
Nanoscale, 4(7), 2378-2386 (2012-03-01)
Semiconductor nanoparticles (NPs) are being actively explored for applications in medical diagnostics and therapy and numerous electronic devices including solar cells. In this paper we demonstrate the influence of the third generation rigid polyphenylenepyridyl dendrimers (PPPDs) of a different architecture

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service