Skip to Content
Merck
All Photos(3)

Key Documents

208256

Sigma-Aldrich

Tin(II) chloride

reagent grade, 98%

Synonym(s):

Stannous chloride

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
SnCl2
CAS Number:
Molecular Weight:
189.62
EC Number:
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:
NACRES:
NA.21
Assay:
98%
grade:
reagent grade
form:
crystalline
powder or flakes

grade

reagent grade

Quality Level

vapor pressure

33 hPa (~429 °C)

Assay

98%

form

crystalline
powder or flakes

pH

2.18 (20 °C)

bp

652 °C (lit.)

mp

246 °C (lit.)

SMILES string

Cl[SnH2]Cl

InChI

1S/2ClH.Sn/h2*1H;/q;;+2/p-2

InChI key

AXZWODMDQAVCJE-UHFFFAOYSA-L

Looking for similar products? Visit Product Comparison Guide

General description

SnCl2 dihydrate reacts with acetic anhydride to yield anhydrous SnCl2. SnCl2 exhibits reducing properties in acidic media. SnCl2 shows superior catalytic action in its molten state. It is a promising raw material for the chemical vapour deposition (CVD) of semiconducting layers. SnCl2 participates in the reduction of aromatic nitro compounds, nitriles, cyanosilyl ethers and organic azides.

Application

Tin(II) chloride (SnCl2) has been used as a starting material to evaluate the thermal behavior of SnCl2 on heating at the rate of 5°Cmin-1 in air. The product undergoes phase transformation on heating at rate of 5°C min-1 in air, it involves an incomplete evaporation followed by oxidation forming SnO2 nanoparticles. SnCl2 has also been used for the fabrication of Sn-containing carbon nano-structures.
Tin(II) chloride may be used:
  • To catalyze the addition of diazo sulfones, diazo phosphine oxides and diazo phosphonates to aldehydes to form β-keto sulfones, β-keto phosphine oxides and β-keto phosphonates, respectively.
  • Along with trityl chloride, to catalyze the aldol reaction of silyl enol ethers with acetals or aldehydes and the Michael reaction of silyl enol ethers with α,β-unsaturated ketones.
  • As a promoter in the allylic amination of allylic alcohols with amines in the presence of palladium catalyst.

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Aquatic Chronic 3 - Eye Dam. 1 - Met. Corr. 1 - Skin Corr. 1B - Skin Sens. 1 - STOT RE 2 Oral - STOT SE 3

Target Organs

Cardio-vascular system, Respiratory system

Storage Class Code

8B - Non-combustible corrosive hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

A possible scalable method for the synthesis of Sn-containing carbon nanostructures.
Kamali AR and Fray DJ.
Materials Today, e38-e48 (2015)
Transformation of molten SnCl2 to SnO2 nano-single crystals.
Kamali AR, et al.
Ceramics International, 40(6), 8533-8538 (2014)
Tin (II) chloride catalyzed addition of diazo sulfones, diazo phosphine oxides, and diazo phosphonates to aldehydes.
Holmquist CR and Roskamp EJ.
Tetrahedron Letters, 33(9), 1131-1134 (1992)
Palladium-catalyzed allylic amination of allylic alcohols with tin (II) chloride and triethylamine.
Masuyama Y, et al.
Chemistry Letters (Jpn), 24(12), 1121-1122 (1995)
An efficient and extremely mild catalyst system, combined use of trityl chloride and tin (II) chloride, in the aldol and Michael reactions.
Mukaiyama T, et al.
Chemistry Letters (Jpn), 16(3), 491-494 (1987)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service