Products may be shipped at a different temperature than the recommended long-term storage temperature. If the product quality is sensitive to short-term exposure to conditions other than the recommended long-term storage, it will be shipped on wet or dry-ice. If the product quality is NOT affected by short-term exposure to conditions other than the recommended long-term storage, it will be shipped at ambient temperature. As shipping routes are configured for minimum transit times, shipping at ambient temperature helps control shipping costs for our customers. For more information, please refer to the Storage and Transport Conditions document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/316/622/storage-transport-conditions-mk.pdf
Y0001805
Pregabalin
European Pharmacopoeia (EP) Reference Standard
Synonym(s):
(S)-3-(Aminomethyl)-5-methylhexanoic acid, CI-1008, PD-144723
About This Item
Recommended Products
grade
pharmaceutical primary standard
API family
pregabalin
manufacturer/tradename
EDQM
drug control
USDEA Schedule I
application(s)
pharmaceutical (small molecule)
format
neat
storage temp.
2-8°C
SMILES string
OC(C[C@@H](CN)CC(C)C)=O
InChI
1S/C8H17NO2/c1-6(2)3-7(5-9)4-8(10)11/h6-7H,3-5,9H2,1-2H3,(H,10,11)/t7-/m0/s1
InChI key
AYXYPKUFHZROOJ-ZETCQYMHSA-N
Gene Information
human ... CACNA1A(773) , CACNA1B(774) , CACNA1C(775) , CACNA1D(776) , CACNA1E(777) , CACNA1F(778) , CACNA1G(8913) , CACNA1H(8912) , CACNA1I(8911) , CACNA1S(779) , CACNA2D1(781) , CACNA2D2(9254) , CACNA2D3(55799) , CACNA2D4(93589) , CACNB1(782) , CACNB2(783) , CACNB3(784) , CACNB4(785) , CACNG1(786) , CACNG2(10369) , CACNG3(10368) , CACNG4(27092) , CACNG5(27091) , CACNG6(59285) , CACNG7(59284) , CACNG8(59283)
Looking for similar products? Visit Product Comparison Guide
Related Categories
General description
For further information and support please go to the website of the issuing Pharmacopoeia.
Application
Biochem/physiol Actions
Packaging
Other Notes
related product
Signal Word
Warning
Hazard Statements
Precautionary Statements
Hazard Classifications
Repr. 2 - STOT SE 3
Target Organs
Central nervous system
Storage Class Code
11 - Combustible Solids
WGK
WGK 1
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Choose from one of the most recent versions:
Certificates of Analysis (COA)
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
-
How is shipping temperature determined? And how is it related to the product storage temperature?
1 answer-
Helpful?
-
-
How can I determine the shelf life / expiration / retest date of this product?
1 answer-
If this product has an expiration or retest date, it will be shown on the Certificate of Analysis (COA, CofA). If there is no retest or expiration date listed on the product's COA, we do not have suitable stability data to determine a shelf life. For these products, the only date on the COA will be the release date; a retest, expiration, or use-by-date will not be displayed.
For all products, we recommend handling per defined conditions as printed in our product literature and website product descriptions. We recommend that products should be routinely inspected by customers to ensure they perform as expected.
For products without retest or expiration dates, our standard warranty of 1 year from the date of shipment is applicable.
For more information, please refer to the Product Dating Information document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/449/386/product-dating-information-mk.pdfHelpful?
-
Active Filters
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service