Y0001712
Polyoxypropylene stearyl ether
European Pharmacopoeia (EP) Reference Standard
About This Item
Recommended Products
grade
pharmaceutical primary standard
API family
polyoxypropylene stearyl ether
manufacturer/tradename
EDQM
application(s)
pharmaceutical (small molecule)
format
neat
storage temp.
2-8°C
SMILES string
O(CC(O)C)CCCCCCCCCCCCCCCCCC
InChI
1S/C21H44O2/c1-3-4-5-6-7-8-9-10-11-12-13-14-15-16-17-18-19-23-20-21(2)22/h21-22H,3-20H2,1-2H3
InChI key
ZQCIPRGNRQXXSK-UHFFFAOYSA-N
General description
For further information and support please go to the website of the issuing Pharmacopoeia.
Application
Packaging
Other Notes
Storage Class Code
10 - Combustible liquids
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Choose from one of the most recent versions:
Certificates of Analysis (COA)
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Active Filters
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service