Skip to Content
Merck
All Photos(1)

Key Documents

Y0001587

Atomoxetine for impurity A identification

European Pharmacopoeia (EP) Reference Standard

Synonym(s):

(R)-Tomoxetine hydrochloride, (R)-N-Methyl-γ-(2-methyl­phenoxy)benzenepropanamine hydrochloride, Atomoxetine hydrochloride

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C17H21NO · HCl
CAS Number:
Molecular Weight:
291.82
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

pharmaceutical primary standard

API family

atomoxetine

manufacturer/tradename

EDQM

application(s)

pharmaceutical (small molecule)

format

neat

storage temp.

2-8°C

SMILES string

CNCC[C@@H](OC1=CC=CC=C1C)C2=CC=CC=C2.[H]Cl

InChI

1S/C17H21NO.ClH/c1-14-8-6-7-11-16(14)19-17(12-13-18-2)15-9-4-3-5-10-15;/h3-11,17-18H,12-13H2,1-2H3;1H/t17-;/m1./s1

InChI key

LUCXVPAZUDVVBT-UNTBIKODSA-N

Gene Information

human ... SLC6A2(6530)

Looking for similar products? Visit Product Comparison Guide

General description

This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the issuing Pharmacopoeia.For further information and support please go to the website of the issuing Pharmacopoeia.

Application

Atomoxetine for impurity A identification EP Reference standard, intended for use in laboratory tests only as specifically prescribed in the European Pharmacopoeia.

Biochem/physiol Actions

Norepinephrine uptake blocker.

Packaging

The product is delivered as supplied by the issuing Pharmacopoeia. For the current unit quantity, please visit the EDQM reference substance catalogue.

Other Notes

Sales restrictions may apply.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Dylan Chou et al.
Experimental neurology, 255, 19-29 (2014-03-04)
Fear conditioning in animals has been used extensively to model clinical anxiety disorders. While individual animals exhibit marked differences in their propensity to undergo fear conditioning, the physiologically relevant mediators have not yet been fully characterized. Here, we demonstrate that
Atomoxetine-induced mydriasis in a child patient.
Kayhan Bahali et al.
European child & adolescent psychiatry, 23(12), 1231-1232 (2013-11-12)
Angie A Kehagia et al.
Brain : a journal of neurology, 137(Pt 7), 1986-1997 (2014-06-05)
Noradrenergic dysfunction may play a significant role in cognition in Parkinson's disease due to the early degeneration of the locus coeruleus. Converging evidence from patient and animal studies points to the role of noradrenaline in dopaminergically insensitive aspects of the
Atomoxetine-induced wake-time teeth clenching and sleep bruxism in a child patient.
Kayhan Bahali et al.
European child & adolescent psychiatry, 23(12), 1233-1235 (2014-08-28)
Addressing the unique issues of student athletes with ADHD.
Adam E Perrin et al.
The Journal of family practice, 63(5), E1-E9 (2014-05-06)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service