Skip to Content
Merck
All Photos(1)

Key Documents

Y0001574

Mesalazine for system suitability

European Pharmacopoeia (EP) Reference Standard

Synonym(s):

5-Aminosalicylic acid, 5-AS, 5-Amino-2-hydroxybenzoic acid, Mesalamine

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
H2NC6H3-2-(OH)CO2H
CAS Number:
Molecular Weight:
153.14
Beilstein:
2090421
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

pharmaceutical primary standard

API family

mesalazine, mesalamine

manufacturer/tradename

EDQM

mp

275-280 °C (dec.) (lit.)

application(s)

pharmaceutical (small molecule)

format

neat

storage temp.

2-8°C

SMILES string

Nc1ccc(O)c(c1)C(O)=O

InChI

1S/C7H7NO3/c8-4-1-2-6(9)5(3-4)7(10)11/h1-3,9H,8H2,(H,10,11)

InChI key

KBOPZPXVLCULAV-UHFFFAOYSA-N

Gene Information

Looking for similar products? Visit Product Comparison Guide

General description

This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the issuing Pharmacopoeia.For further information and support please go to the website of the issuing Pharmacopoeia.

Application

5-Aminosalicylic acid is a peroxidase substrate suitable for use in ELISA procedures. This substrate produces a soluble end product that is brown in color and can be read spectrophotometrically at 450 nm. The reaction may be stopped with 3 N NaOH and read at 550 nm.

Packaging

The product is delivered as supplied by the issuing Pharmacopoeia. For the current unit quantity, please visit the EDQM reference substance catalogue.

Other Notes

Sales restrictions may apply.

related product

Product No.
Description
Pricing

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - Skin Sens. 1 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

No data available

Flash Point(C)

No data available


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

A H Steinhart et al.
Alimentary pharmacology & therapeutics, 25(12), 1389-1399 (2007-06-02)
To evaluate the effectiveness of pH 6-/pH 7-dependent and controlled-release mesalazines in maintaining medically and surgically induced Crohn's disease remission. A systematic search identified 13 randomized controlled trials (RCTs). The rate of symptomatic relapse (Crohn's disease activity index >150, or
P D R Higgins et al.
Alimentary pharmacology & therapeutics, 29(3), 247-257 (2008-10-24)
Ulcerative colitis (UC) can be maintained in remission with 5-aminosalicylic acid (5-ASA) medications, but frequent non-adherence by patients who are feeling well has been associated with more frequent flares of colitis. To perform a systematic review of the published literature
Marietta Iacucci et al.
Canadian journal of gastroenterology = Journal canadien de gastroenterologie, 24(2), 127-133 (2010-02-13)
5-aminosalicylic acid (5-ASA) preparations (eg, mesalazine, mesalamine) are well-established preparations used in the management of inflammatory bowel disease. These drugs are most useful for the treatment of mild to moderate flares of ulcerative colitis and, especially, for maintenance of remission.
Altamash I Qureshi et al.
Advanced drug delivery reviews, 57(2), 281-302 (2004-11-24)
Sulfasalazine's role as the first-line of therapy in patients with inflammatory bowel disease has led to the development of other "designer" aminosalicylates, which eliminate the sulfa-moiety, and attempt to target the topically active mesalamine to the inflamed bowel. Olsalazine sodium
Alexander C Ford et al.
The American journal of gastroenterology, 106(12), 2070-2077 (2011-09-07)
Maintenance therapy with 5-aminosalicylates (5-ASAs) is recommended in patients with quiescent ulcerative colitis (UC), but compliance rates are low. Once-daily dosing may improve adherence, but impact on the relapse of disease activity is unclear as no previous meta-analysis has studied

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service