Skip to Content
Merck
All Photos(1)

Documents

Y0001409

Testosterone propionate for system suitability

European Pharmacopoeia (EP) Reference Standard

Synonym(s):

Testosterone propionate, 17β-Hydroxy-4-androsten-3-one 17-propionate, 17β-Propionyloxy-4-androsten-3-one, 4-Androsten-17β-ol-3-one 17-propionate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C22H32O3
CAS Number:
Molecular Weight:
344.49
Beilstein:
3221760
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

pharmaceutical primary standard

API family

testosterone

manufacturer/tradename

EDQM

drug control

regulated under CDSA - not available from Sigma-Aldrich Canada

application(s)

pharmaceutical (small molecule)

format

neat

storage temp.

2-8°C

SMILES string

[H][C@@]12CCC3=CC(=O)CC[C@]3(C)[C@@]1([H])CC[C@]4(C)[C@H](CC[C@@]24[H])OC(=O)CC

InChI

1S/C22H32O3/c1-4-20(24)25-19-8-7-17-16-6-5-14-13-15(23)9-11-21(14,2)18(16)10-12-22(17,19)3/h13,16-19H,4-12H2,1-3H3/t16-,17-,18-,19-,21-,22-/m0/s1

InChI key

PDMMFKSKQVNJMI-BLQWBTBKSA-N

Gene Information

human ... AR(367)

Looking for similar products? Visit Product Comparison Guide

General description

This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the issuing Pharmacopoeia.For further information and support please go to the website of the issuing Pharmacopoeia.

Application

Testosterone propionate for system suitability EP Reference standard, intended for use in laboratory tests only as specifically prescribed in the European Pharmacopoeia.

Packaging

The product is delivered as supplied by the issuing Pharmacopoeia. For the current unit quantity, please visit the EDQM reference substance catalogue.

Other Notes

Sales restrictions may apply.

related product

Product No.
Description
Pricing

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Acute 1 - Carc. 2 - Repr. 1A

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Kyung Pyo Kang et al.
Molecular medicine reports, 9(6), 2061-2068 (2014-04-01)
Inflammation is a key mediator of renal ischemia-reperfusion (IR) injury. Gender disparities have been reported in acute and chronic kidney disease. In particular, males are considered to be more susceptible to renal ischemic injury compared with females according to animal
Lei Wan et al.
Cancer prevention research (Philadelphia, Pa.), 7(12), 1228-1239 (2014-10-16)
Consumption of tomato products containing the carotenoid lycopene is associated with a reduced risk of prostate cancer. To identify gene expression patterns associated with early testosterone-driven prostate carcinogenesis, which are impacted by dietary tomato and lycopene, wild-type (WT) and transgenic
A B Firdous et al.
Food & function, 5(10), 2632-2645 (2014-08-29)
Prostate cancer incidence and mortality rates have increased over the past years. The purpose of the present study was to examine the molecular mechanism underlying the chemopreventive effects of quercetin on prostate cancer in an in vivo model. Sprague-Dawley male
E M Beckett et al.
Reproduction (Cambridge, England), 148(2), 199-209 (2014-05-21)
Gestational testosterone treatment causes maternal hyperinsulinemia, intrauterine growth retardation (IUGR), low birth weight, and adult reproductive and metabolic dysfunctions. Sheep models of IUGR demonstrate placental insufficiency as an underlying cause of IUGR. Placental compromise is probably the cause of fetal
Mahmoud M Said et al.
Journal of toxicology and environmental health. Part A, 78(7), 453-465 (2015-03-19)
Benign prostatic hyperplasia (BPH), a disease occurring frequently among elderly males, is a slow progressive enlargement of the fibromuscular and epithelial structures of the prostate gland. Dietary factors may influence the prostate and exert an influence on prostatic growth and

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service