Skip to Content
Merck
All Photos(1)

Key Documents

Y0001294

Iodixanol

European Pharmacopoeia (EP) Reference Standard

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C35H44I6N6O15
CAS Number:
Molecular Weight:
1550.18
UNSPSC Code:
41116107
NACRES:
NA.24
Pricing and availability is not currently available.

grade

pharmaceutical primary standard

API family

iodixanol

manufacturer/tradename

EDQM

application(s)

pharmaceutical (small molecule)

format

neat

storage temp.

2-8°C

SMILES string

Ic1c(c(c(c(c1C(=O)NCC(O)CO)I)C(=O)NCC(O)CO)I)N(CC(O)CN(c2c(c(c(c(c2I)C(=O)NCC(O)CO)I)C(=O)NCC(O)CO)I)C(=O)C)C(=O)C

InChI

1S/C35H44I6N6O15/c1-13(52)46(30-26(38)20(32(59)42-3-15(54)9-48)24(36)21(27(30)39)33(60)43-4-16(55)10-49)7-19(58)8-47(14(2)53)31-28(40)22(34(61)44-5-17(56)11-50)25(37)23(29(31)41)35(62)45-6-18(57)12-51/h15-19,48-51,54-58H,3-12H2,1-2H3,(H,42,59)(H,43,60)(H,44,61)(H,45,62)

InChI key

NBQNWMBBSKPBAY-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the issuing Pharmacopoeia.For further information and support please go to the website of the issuing Pharmacopoeia.

Application

Iodixanol EP Reference standard, intended for use in laboratory tests only as specifically prescribed in the European Pharmacopoeia.

Packaging

The product is delivered as supplied by the issuing Pharmacopoeia. For the current unit quantity, please visit the EDQM reference substance catalogue.

Other Notes

Sales restrictions may apply.

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Sonja M Kandel et al.
Radiology, 270(1), 125-130 (2013-09-14)
To analyze the effect of the duration of contrast material bolus injection on perfusion values in a swine model by using the maximum slope method. This study was approved by the institutional animal care committee. Twenty pigs (weight range, 63-77
Hyun-Jung Jang et al.
AJR. American journal of roentgenology, 201(2), 314-321 (2013-07-26)
The purpose of this study was to identify the essential number of phases from multiphasic CT for 1- to 2-cm hepatocellular carcinoma (HCC) on surveillance ultrasound and to compare the results with the American Association for the Study of Liver
Kartik Jhaveri et al.
AJR. American journal of roentgenology, 201(2), W292-W296 (2013-07-26)
The purpose of this study is to evaluate the interval growth, tumor recurrence, and metastatic disease occurrence of cystic renal cell carcinoma (RCC). Pre-and posttreatment imaging of 47 histologically proven cystic RCCs, with at least 6 months of pretreatment imaging
Celso Henrique Alves et al.
Human molecular genetics, 23(13), 3384-3401 (2014-02-05)
In humans, the Crumbs homolog-1 (CRB1) gene is mutated in autosomal recessive Leber congenital amaurosis and early-onset retinitis pigmentosa. In mammals, the Crumbs family is composed of: CRB1, CRB2, CRB3A and CRB3B. Recently, we showed that removal of mouse Crb2
Michael Reed et al.
JACC. Cardiovascular interventions, 2(7), 645-654 (2009-07-25)
We sought to compare the nephrotoxicity of the iso-osmolar contrast medium, iodixanol, to low-osmolar contrast media (LOCM). Contrast-induced acute kidney injury (CI-AKI) is a common cause of in-hospital renal failure. A prior meta-analysis suggested that iodixanol (Visipaque, GE Healthcare, Princeton

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service