Skip to Content
Merck
All Photos(1)

Key Documents

Y0001285

Sertraline impurity E

European Pharmacopoeia (EP) Reference Standard

Synonym(s):

(R)-(−)-Mandelic acid, (R)-α-Hydroxyphenylacetic acid

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C6H5CH(OH)CO2H
CAS Number:
Molecular Weight:
152.15
Beilstein:
2691094
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

pharmaceutical primary standard

API family

sertraline

manufacturer/tradename

EDQM

mp

131-133 °C (lit.)

application(s)

pharmaceutical (small molecule)

format

neat

storage temp.

2-8°C

SMILES string

O[C@@H](C(O)=O)c1ccccc1

InChI

1S/C8H8O3/c9-7(8(10)11)6-4-2-1-3-5-6/h1-5,7,9H,(H,10,11)/t7-/m1/s1

InChI key

IWYDHOAUDWTVEP-SSDOTTSWSA-N

Looking for similar products? Visit Product Comparison Guide

General description

This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the Issuing Pharmacopoeia. For further information and support please go to the website of the issuing Pharmacopoeia.

Application

Sertraline impurity E EP Reference standard, intended for use in laboratory tests only as specifically prescribed in the European Pharmacopoeia.

Packaging

The product is delivered as supplied by the issuing Pharmacopoeia. For the current unit quantity, please visit the EDQM reference substance catalogue.

Other Notes

Sales restrictions may apply.

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Precautionary Statements

Hazard Classifications

Eye Dam. 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Selahattin Bozkurt et al.
Chirality, 24(2), 129-136 (2011-12-20)
Novel chiral calix[4]arene derivatives bearing amino alcohol moieties at the lower rim have been synthesized from the reaction of p-tert-butylcalix[4]arene diester with various amino alcohols. The transport of amino acid esters (phenylglycine, phenylalanine, and tryptophan methyl esters hydrochloride) and mandelic
M Monier et al.
International journal of biological macromolecules, 55, 207-213 (2013-01-30)
An enantioselective S-mandelic acid (S-MA) imprinted chitosan (SMIC) was prepared by cross-linking of chitosan using formaldehyde cross-linker, in the presence of S-MA as an imprint template molecule and 0.5% acetic acid solution as a solvent. Non-imprinted cross-linked chitosan (NIC) as
Chao Ma et al.
Chirality, 23(5), 379-382 (2011-04-14)
This work reports the chiral separation of D,L-mandelic acid with cellulose membranes. Cellulose was chosen as membrane material because it possesses multichiral carbon atoms in its molecular structure unit. The flux and permselective properties of membrane using aqueous solutions of
Liang Jin et al.
Zeitschrift fur Naturforschung. C, Journal of biosciences, 66(9-10), 499-506 (2011-12-24)
To study the effects of mandelic acid (MA) on the brown planthopper (BPH), Nilaparvata lugens, the survival rate and behaviour of BPH fed on an artificial diet with different dosages of MA was observed. The survival rate of BPH decreased
Yubo Wu et al.
The Journal of organic chemistry, 76(14), 5685-5695 (2011-05-31)
We have prepared chiral fluorescent bisboronic acid sensors with 3,6-dithiophen-2-yl-9H-carbazole as the fluorophore. The thiophene moiety was used to extend the π-conjugation framework of the fluorophore in order to red-shift the fluorescence emission and, at the same time, to enhance

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service