Skip to Content
Merck
All Photos(1)

Key Documents

Y0001225

Valaciclovir for system suitability

European Pharmacopoeia (EP) Reference Standard

Synonym(s):

Valacyclovir hydrochloride

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C13H20N6O4 · HCl
CAS Number:
Molecular Weight:
360.80
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

pharmaceutical primary standard

API family

valacyclovir

manufacturer/tradename

EDQM

application(s)

pharmaceutical (small molecule)

format

neat

storage temp.

2-8°C

SMILES string

Cl.CC(C)[C@H](N)C(=O)OCCOCn1cnc2C(=O)NC(N)=Nc12

InChI

1S/C13H20N6O4.ClH/c1-7(2)8(14)12(21)23-4-3-22-6-19-5-16-9-10(19)17-13(15)18-11(9)20;/h5,7-8H,3-4,6,14H2,1-2H3,(H3,15,17,18,20);1H/t8-;/m0./s1

InChI key

ZCDDBUOENGJMLV-QRPNPIFTSA-N

Looking for similar products? Visit Product Comparison Guide

General description

This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the issuing Pharmacopoeia.
For further information and support please go to the website of the issuing Pharmacopoeia.
Valacyclovir is a prodrug of acyclovir. It is an L-valyl ester and works against herpes simplex virus type and varicella zoster virus.

Application

This European Pharmacopoeia reference standard is intended for use only as specifically prescribed in the European Pharmacopoeia.

Packaging

The product is delivered as supplied by the issuing Pharmacopoeia. For the current unit quantity, please visit the EDQM reference substance catalogue.

Other Notes

Sales restrictions may apply.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Acute Tox. 4 Oral

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

UV spectrophotometric method for the estimation of valacyclovir hcl in tablet dosage form.
Ganesh, M., et al.
Journal of Chemistry, 6.3, 814-818 (2009)
Selective and rapid liquid chromatography/negative-ion electrospray ionization mass spectrometry method for the quantification of valacyclovir and its metabolite in human plasma.
Kasiari M
Journal of Chromatography. B, Analytical Technologies in the Biomedical and Life Sciences, 864(1-2), 78-86 (2008)
Mary J Burton et al.
The American journal of the medical sciences, 348(6), 455-459 (2014-08-28)
We performed a pilot study examining the safety and tolerability of valacyclovir in veterans with herpes simplex virus type 2 and hepatitis C virus (HCV) coinfection. We performed a randomized double-blind, placebo-controlled, crossover clinical trial in U.S. veterans with genotype
Laurence Le Cleach et al.
The Cochrane database of systematic reviews, 8(8), CD009036-CD009036 (2014-08-05)
Genital herpes is caused by herpes simplex virus 1 (HSV-1) or 2 (HSV-2). Some infected people experience outbreaks of genital herpes, typically, characterized by vesicular and erosive localized painful genital lesions. To compare the effectiveness and safety of three oral
Shlomit Schaal et al.
JAMA ophthalmology, 132(7), 881-882 (2014-04-20)
Acute retinal necrosis (ARN) is an infectious retinitis primarily caused by the herpesviruses. Although the Epstein-Barr virus (EBV) has been implicated as a cause of ARN, to our knowledge, there has been no histopathologic documentation. We report the clinical history

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service