Skip to Content
Merck
All Photos(1)

Key Documents

Y0001121

Adapalene

European Pharmacopoeia (EP) Reference Standard

Synonym(s):

6-(4-Methoxy-3-tricyclo[3.3.1.13,7]dec-1-ylphenyl)-2-naphthalenecarboxylic acid, 6-[3-(1-Adamantyl)-4-methoxyphenyl]-2-naphthoic acid, CD-271, Differin

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C28H28O3
CAS Number:
Molecular Weight:
412.52
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

pharmaceutical primary standard

API family

adapalene

manufacturer/tradename

EDQM

application(s)

pharmaceutical (small molecule)

format

neat

storage temp.

2-8°C

SMILES string

COc1ccc(cc1C23C[C@H]4C[C@H](C[C@H](C4)C2)C3)-c5ccc6cc(ccc6c5)C(O)=O

InChI

1S/C28H28O3/c1-31-26-7-6-23(21-2-3-22-12-24(27(29)30)5-4-20(22)11-21)13-25(26)28-14-17-8-18(15-28)10-19(9-17)16-28/h2-7,11-13,17-19H,8-10,14-16H2,1H3,(H,29,30)/t17-,18+,19-,28-

InChI key

LZCDAPDGXCYOEH-AADAIPAGSA-N

Looking for similar products? Visit Product Comparison Guide

General description

This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the Issuing Pharmacopoeia. For further information and support please go to the website of the issuing Pharmacopoeia.

Application

Adapalene EP Reference standard, intended for use in laboratory tests only as specifically prescribed in the European Pharmacopoeia.

Biochem/physiol Actions

Retinoic acid analogue that is a RARβ and RARγ agonist (AC50 values are 2.2, 9.3, 22 and > 1000 nM for RARβ, RARγ, RARα and RXRα receptors respectively). Inhibits proliferation and induces apoptosis in colorectal cancer cells in vitro. Displays comedolytic activity. Its unique pharmacological properties make it superior to other retinoids for the treatment of acne.

Packaging

The product is delivered as supplied by the issuing Pharmacopoeia. For the current unit quantity, please visit the EDQM reference substance catalogue.

Other Notes

Sales restrictions may apply.

related product

Pictograms

Health hazard

Signal Word

Warning

Hazard Statements

Hazard Classifications

Repr. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

John Waugh et al.
American journal of clinical dermatology, 5(5), 369-371 (2004-11-24)
Adapalene (Differin) is a retinoid agent indicated for the topical treatment of acne vulgaris. In clinical trials, 0.1% adapalene gel has proved to be effective in this indication and was as effective as 0.025% tretinoin gel, 0.1% tretinoin microsphere gel
Adapalene for the treatment of acne vulgaris.
J S Weiss et al.
Journal of the American Academy of Dermatology, 39(2 Pt 3), S50-S54 (1998-08-14)
B Shroot et al.
Journal of the American Academy of Dermatology, 36(6 Pt 2), S96-103 (1997-06-01)
Retinoid research in the field of dermatology has been influenced by the clinical success of topical tretinoin and oral isotretinoin in the treatment of acne, and by the discovery of high-affinity binding proteins for retinoic acid mediating its action and
J E Wolf
Journal of the European Academy of Dermatology and Venereology : JEADV, 15 Suppl 3, 23-29 (2002-02-15)
Topical retinoids are a mainstay in the treatment of acne vulgaris. In the past these agents have been associated with irritant effects, however, newer generations of topical retinoids have emerged that have been designed to be less irritating. This paper
R N Brogden et al.
Drugs, 53(3), 511-519 (1997-03-01)
Adapalene, a naphthoic acid derivative with retinoid-like activity, is used for the topical treatment of mild to moderate acne vulgaris. It binds to retinoic acid receptors found predominantly in the terminal differentiation zone of epidermis and is more active than

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service