Skip to Content
Merck
All Photos(1)

Documents

Y0000902

Racecadotril for peak identification

European Pharmacopoeia (EP) Reference Standard

Synonym(s):

Racecadotril, (RS)-Benzyl N-[3-(acetylthio)-2-benzylpropanoyl]glycinate, (±)-Acetorphan, Acetorphan, Cadotril, Redotil, Tiorfan, Zedott

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C21H23NO4S
CAS Number:
Molecular Weight:
385.48
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

pharmaceutical primary standard

API family

racecadotril

manufacturer/tradename

EDQM

application(s)

pharmaceutical (small molecule)

format

neat

storage temp.

2-8°C

SMILES string

CC(=O)SCC(Cc1ccccc1)C(=O)NCC(=O)OCc2ccccc2

InChI

1S/C21H23NO4S/c1-16(23)27-15-19(12-17-8-4-2-5-9-17)21(25)22-13-20(24)26-14-18-10-6-3-7-11-18/h2-11,19H,12-15H2,1H3,(H,22,25)

InChI key

ODUOJXZPIYUATO-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the issuing Pharmacopoeia.For further information and support please go to the website of the issuing Pharmacopoeia.

Application

Racecadotril for peak identification EP Reference standard, intended for use in laboratory tests only as specifically prescribed in the European Pharmacopoeia.

Biochem/physiol Actions

Racecadotril is a neutral endopeptidase inhibitor, an antidiarrheal drug which acts as a peripherally acting enkephalinase inhibitor.

Packaging

The product is delivered as supplied by the issuing Pharmacopoeia. For the current unit quantity, please visit the EDQM reference substance catalogue.

Other Notes

Sales restrictions may apply.

Pictograms

Exclamation markEnvironment

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Acute 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Ramón Tormo et al.
Acta paediatrica (Oslo, Norway : 1992), 97(8), 1008-1015 (2008-05-09)
In developing countries acute infectious diarrhoea remains one of the leading causes of death among young children, especially those under 1 year of age. In contrast, in industrialized nations the death rate is very low, although the disease is an
J I Emparanza Knörr et al.
Anales de pediatria (Barcelona, Spain : 2003), 69(5), 432-438 (2009-01-09)
To estimate, through a systematic review of the literature, the efficacy of racecadotril in the treatment of acute diarrhoea. Randomised trials carried out in children comparing racecadotril with placebo in terms of diarrhoea recovery, stools output and adverse effects were
J C Schwartz
International journal of antimicrobial agents, 14(1), 75-79 (2000-03-16)
Since enkephalins were discovered in 1975, it has become clear that they play an antisecretory role in the gastrointestinal tract. Hence a rational research programme was directed at the development of a drug that would preserve these neurotransmitter peptides in
H Szajewska et al.
Alimentary pharmacology & therapeutics, 26(6), 807-813 (2007-09-05)
Racecadotril (acetorphan) is an antisecretory drug that exerts its antidiarrhoeal effects by inhibiting intestinal enkephalinase. To summarize studies testing the efficacy and safety of racecadotril for treating children with acute gastroenteritis. Reports were gathered by searching electronic databases MEDLINE, EMBASE
S Huighebaert et al.
Digestive diseases and sciences, 48(2), 239-250 (2003-03-20)
Racecadotril is an enkephalinase inhibitor, presented as a purely antisecretory agent with advantages over the opiate-receptor agonist loperamide in the treatment of diarrhea. A critical review of the literature and the models used was performed. Although pretreatment with high doses

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service