Skip to Content
Merck
All Photos(1)

Key Documents

Y0000821

Molsidomine

European Pharmacopoeia (EP) Reference Standard

Synonym(s):

N-(Ethoxycarbonyl)-3-(4-morpholino)sydnone imine, SIN-10

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C9H14N4O4
CAS Number:
Molecular Weight:
242.23
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

pharmaceutical primary standard

API family

molsidomine

manufacturer/tradename

EDQM

application(s)

pharmaceutical (small molecule)

format

neat

SMILES string

CCOC(=O)N=C1O[N-][N+](=C1)N2CCOCC2

InChI

1S/C9H14N4O4/c1-2-16-9(14)10-8-7-13(11-17-8)12-3-5-15-6-4-12/h7H,2-6H2,1H3/b10-8-

InChI key

XLFWDASMENKTKL-NTMALXAHSA-N

Looking for similar products? Visit Product Comparison Guide

General description

This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the issuing Pharmacopoeia.For further information and support please go to the website of the issuing Pharmacopoeia.

Application

Molsidomine EP Reference standard, intended for use in laboratory tests only as specifically prescribed in the European Pharmacopoeia.

Packaging

The product is delivered as supplied by the issuing Pharmacopoeia. For the current unit quantity, please visit the EDQM reference substance catalogue.

Other Notes

Sales restrictions may apply.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Acute Tox. 4 Oral

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

R E Nitz et al.
Pharmacotherapy, 7(1), 28-37 (1987-01-01)
The long-acting antianginal drug molsidomine has been shown experimentally to reduce myocardial infarct size when administered prior to or after cardiac insult. This is due to several drug actions. Dilation of postcapillary capacitance vessels diminishes venous return, preload, heart dimensions
[Molsidomine (Corvaton)].
O M Eliseev
Terapevticheskii arkhiv, 66(9), 82-88 (1994-01-01)
R E Nitz et al.
American heart journal, 109(3 Pt 2), 631-636 (1985-03-01)
The cardioprotective and antithrombotic activity of molsidomine, a novel therapeutic agent for the treatment of coronary heart disease, was investigated in a series of animal models of myocardial ischemia. Molsidomine given to dogs with marked ST segment elevation (epicardial electrogram)
[Pharmacology of molsidomine and its active metabolites].
E Böhme
Medizinische Klinik (Munich, Germany : 1983), 85 Suppl 1, 7-10 (1990-02-01)
[Molsidomine in the treatment of ischemic heart disease].
S Sterkowicz
Wiadomosci lekarskie (Warsaw, Poland : 1960), 40(20), 1389-1395 (1987-10-15)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service