Skip to Content
Merck
All Photos(1)

Key Documents

Y0000775

Benzbromarone

European Pharmacopoeia (EP) Reference Standard

Synonym(s):

3-(3,5-Dibromo-4-hydroxybenzoyl)-2-ethylbenzofuran

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C17H12Br2O3
CAS Number:
Molecular Weight:
424.08
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

pharmaceutical primary standard

API family

benzbromarone

manufacturer/tradename

EDQM

application(s)

pharmaceutical (small molecule)

format

neat

storage temp.

2-8°C

SMILES string

CCc1oc2ccccc2c1C(=O)c3cc(Br)c(O)c(Br)c3

InChI

1S/C17H12Br2O3/c1-2-13-15(10-5-3-4-6-14(10)22-13)16(20)9-7-11(18)17(21)12(19)8-9/h3-8,21H,2H2,1H3

InChI key

WHQCHUCQKNIQEC-UHFFFAOYSA-N

Gene Information

Looking for similar products? Visit Product Comparison Guide

General description

This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the Issuing Pharmacopoeia. For further information and support please go to the website of the issuing Pharmacopoeia.

Application

Benzbromarone EP Reference standard, intended for use in laboratory tests only as specifically prescribed in the European Pharmacopoeia.

Packaging

The product is delivered as supplied by the issuing Pharmacopoeia. For the current unit quantity, please visit the EDQM reference substance catalogue.

Other Notes

Sales restrictions may apply.

Pictograms

Skull and crossbones

Signal Word

Danger

Hazard Statements

Precautionary Statements

Hazard Classifications

Acute Tox. 3 Oral

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Dingyu Wang et al.
Cardiovascular toxicology, 21(3), 192-205 (2020-10-26)
High levels of serum uric acid is closely associated with atrial fibrillation (AF); nonetheless, the detailed mechanisms remain unknown. Therefore, this work examined the intricate mechanisms of AF triggered by hyperuricemia and the impact of the uricosuric agent benzbromarone on
Ming-Han H Lee et al.
Drug safety, 31(8), 643-665 (2008-07-19)
Benzbromarone, a potent uricosuric drug, was introduced in the 1970s and was viewed as having few associated serious adverse reactions. It was registered in about 20 countries throughout Asia, South America and Europe. In 2003, the drug was withdrawn by
Kaoru Kobayashi et al.
Biopharmaceutics & drug disposition, 33(8), 466-473 (2012-08-31)
Benzbromarone (BBR) is metabolized to 1'-hydroxy BBR and 6-hydroxy BBR in the liver. 6-Hydroxy BBR is further metabolized to 5,6-dihydroxy BBR. The aim of this study was to identify the CYP isozymes involved in the metabolism of BBR to 1'-hydroxy
Zhijian Lin et al.
Journal of medicinal food, 17(11), 1214-1221 (2014-10-15)
Inulin, a group of dietary fibers, is reported to improve the metabolic disorders. In the present study, we investigated the effects of chicory inulin on serum metabolites of uric acid (UA), lipids, glucose, and abdominal fat deposition in quail model
Tip W Loo et al.
Biochemistry, 50(21), 4393-4395 (2011-04-28)
Deletion of Phe508 from the first nucleotide-binding domain of the CFTR chloride channel causes cystic fibrosis because it inhibits protein folding. Indirect approaches such as incubation at low temperatures can partially rescue ΔF508 CFTR, but the protein is unstable at

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service