Skip to Content
Merck
All Photos(1)

Key Documents

Y0000735

Dacarbazine impurity B

European Pharmacopoeia (EP) Reference Standard

Synonym(s):

5-Amino-4-imidazolecarboxamide

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C4H6N4O
CAS Number:
Molecular Weight:
126.12
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

pharmaceutical primary standard

API family

dacarbazine

manufacturer/tradename

EDQM

mp

164-170 °C (lit.)

application(s)

pharmaceutical (small molecule)

format

neat

storage temp.

2-8°C

SMILES string

NC(=O)c1nc[nH]c1N

InChI

1S/C4H6N4O/c5-3-2(4(6)9)7-1-8-3/h1H,5H2,(H2,6,9)(H,7,8)

InChI key

DVNYTAVYBRSTGK-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the issuing Pharmacopoeia.For further information and support please go to the website of the issuing Pharmacopoeia.

Application

Dacarbazine impurity B EP Reference standard, intended for use in laboratory tests only as specifically prescribed in the European Pharmacopoeia.

Packaging

The product is delivered as supplied by the issuing Pharmacopoeia. For the current unit quantity, please visit the EDQM reference substance catalogue.

Other Notes

Sales restrictions may apply.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Takashi Sasaki et al.
American journal of physiology. Endocrinology and metabolism, 306(9), E1085-E1092 (2014-03-20)
Exercise can effectively ameliorate type 2 diabetes and insulin resistance. Here we show that the mRNA levels of one of peroxisome proliferator-activated receptor (PPAR) family members, PPARγ1, and genes related to energy metabolism, including PPARγ coactivator-1 protein-1α (PGC-1α) and lipoprotein
Xiaodong Zhang et al.
American journal of physiology. Endocrinology and metabolism, 307(11), E1047-E1056 (2014-10-16)
Fat-specific protein 27 (FSP27) plays a pivotal role in controlling the formation of large lipid droplet and energy metabolism. The cellular levels of FSP27 are tightly regulated through the proteasomal ubiquitin-mediated degradation. However, the upstream signals that trigger FSP27 degradation
Melissa M Thomas et al.
FASEB journal : official publication of the Federation of American Societies for Experimental Biology, 28(5), 2098-2107 (2014-02-14)
AMP-activated protein kinase (AMPK) is a master regulator of metabolism. While muscle-specific AMPK β1β2 double-knockout (β1β2M-KO) mice display alterations in metabolic and mitochondrial capacity, their severe exercise intolerance suggested a secondary contributor to the observed phenotype. We find that tibialis
Adam J Coggins et al.
Organic & biomolecular chemistry, 13(11), 3378-3381 (2015-02-07)
We report an efficient, atom economical general acid-base catalyzed one-step multi-gram synthesis of azepinomycin from commercially available compounds in water. We propose that the described pH-dependent Amadori rearrangement, which couples an amino-imidazole and simple sugar, is of importance as a
Nicola E Clarke et al.
Clinical science (London, England : 1979), 126(7), 507-516 (2013-10-24)
ACE2 (angiotensin-converting enzyme 2) counterbalances the actions of ACE (angiotensin-converting enzyme) by metabolizing its catalytic product, the vasoactive and fibrogenic peptide AngII (angiotensin II), into Ang-(1-7) [angiotensin-(1-7)]. Enhanced ACE2 expression may be protective in diabetes, cardiovascular disease and cancer. However

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service