Skip to Content
Merck
All Photos(1)

Key Documents

Y0000612

Dopexamine dihydrochloride

European Pharmacopoeia (EP) Reference Standard

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C22H32N2O2 · 2HCl
CAS Number:
Molecular Weight:
429.42
UNSPSC Code:
41116107
NACRES:
NA.24

grade

pharmaceutical primary standard

API family

dopexamine

manufacturer/tradename

EDQM

application(s)

pharmaceutical (small molecule)

format

neat

storage temp.

2-8°C

SMILES string

[Cl-].[Cl-].N(CCCCCCNCCc2ccccc2)CCc1cc(c(cc1)O)O.[H+].[H+]

InChI

1S/C22H32N2O2.2ClH/c25-21-11-10-20(18-22(21)26)13-17-24-15-7-2-1-6-14-23-16-12-19-8-4-3-5-9-19;;/h3-5,8-11,18,23-26H,1-2,6-7,12-17H2;2*1H

InChI key

VPDULUNRSQWWJB-UHFFFAOYSA-N

General description

This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the issuing Pharmacopoeia.For further information and support please go to the website of the issuing Pharmacopoeia.

Application

Dopexamine dihydrochloride EP Reference standard, intended for use in laboratory tests only as specifically prescribed in the European Pharmacopoeia.

Packaging

The product is delivered as supplied by the issuing Pharmacopoeia. For the current unit quantity, please visit the EDQM reference substance catalogue.

Other Notes

Sales restrictions may apply.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Shaman Jhanji et al.
Critical care (London, England), 14(4), R151-R151 (2010-08-12)
Post-operative outcomes may be improved by the use of flow related end-points for intra-venous fluid and/or low dose inotropic therapy. The mechanisms underlying this benefit remain uncertain. The objective of this study was to assess the effects of stroke volume
N E El Mokhtari et al.
European journal of medical research, 13(10), 459-463 (2008-11-15)
To examine the effects of a therapy with dopexamine/dopamine in comparison with a regimen of dobutamine/dopamine on the outcome of patients with profound cardiogenic shock. Twenty patients presenting with an acute cardiogenic shock assisted with mechanical ventilation, being refractory to
Nour Eddine El Mokhtari et al.
European journal of medical research, 12(11), 563-567 (2007-11-21)
To examine the effects of a therapy with dopexamine/dopamine in comparison with a regimen of dobutamine/dopamine on the outcome of patients with profound cardiogenic shock. Twenty patients presenting with an acute cardiogenic shock assisted with mechanical ventilation, being refractory to
Meta-analyses of the effects of dopexamine in major surgery: do all roads lead to Rome?
J J Pandit
Anaesthesia, 64(6), 585-588 (2009-05-21)
N Mayeur et al.
Annales francaises d'anesthesie et de reanimation, 29(11), 759-764 (2010-10-12)
To evaluate the 6 hours haemodynamic effects of dopexamine (DPX) infusion in septic shock patients with persistent hyperlactatemia treated with high dose of norepinephrine (NE). Preliminary, prospective, uncontrolled study. Twenty-one septic shock with NE>0.5 μg/kg/min, venous mixed oxygen saturation (ScvO(2)/SvO(2))>70%

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service