Skip to Content
Merck
All Photos(1)

Key Documents

Y0000610

Cladribine impurity C

European Pharmacopoeia (EP) Reference Standard

Synonym(s):

2-Chloroadenine, 2-Chloro-6-aminopurine

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C5H4ClN5
CAS Number:
Molecular Weight:
169.57
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

pharmaceutical primary standard

API family

cladribine

manufacturer/tradename

EDQM

mp

384-389 °C

application(s)

pharmaceutical (small molecule)

format

neat

shipped in

wet ice

storage temp.

−20°C

SMILES string

Nc1nc(Cl)nc2nc[nH]c12

InChI

1S/C5H4ClN5/c6-5-10-3(7)2-4(11-5)9-1-8-2/h1H,(H3,7,8,9,10,11)

InChI key

HBJGQJWNMZDFKL-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the Issuing Pharmacopoeia. For further information and support please go to the website of the issuing Pharmacopoeia.

Application

Cladribine impurity C EP Reference standard, intended for use in laboratory tests only as specifically prescribed in the European Pharmacopoeia.

Packaging

The product is delivered as supplied by the issuing Pharmacopoeia. For the current unit quantity, please visit the EDQM reference substance catalogue.

Other Notes

Sales restrictions may apply.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Laura Willems et al.
Journal of molecular and cellular cardiology, 38(2), 245-256 (2005-02-09)
Aging is associated with reduced tolerance to ischemic insult, and genesis of this intolerant phenotype is poorly understood. We characterized effects of aging and gender on cardiovascular function and cell damage during 20 min ischemia and 60 min reperfusion in
P Hentosh et al.
Nucleic acids research, 19(11), 3143-3148 (1991-06-11)
The purine analog, 2-chloro-2'-deoxyadenosine triphosphate (CldATP), was incorporated enzymatically in place of dATP into the minus strand of M13mp18 duplex DNA. Its effect on protein-DNA interactions was assessed by determining the amount of DNA cleavage by type II restriction endonucleases.
Kinga Banach et al.
Nucleosides, nucleotides & nucleic acids, 24(5-7), 465-469 (2005-10-27)
A new kinetic model is presented for analysis of experimental data of oxidation process catalyzed by milk xanthine oxidase. The kinetics for two substrates, xanthine and its analog 2-chloroadenine, in a broad pH range (5.8-9.0) are best described by an
Peng Li et al.
Biochemical pharmacology, 79(7), 1015-1024 (2009-11-10)
Since the heterologous tolerance that develops after chronic morphine administration has been proposed to be an adaptive process, it follows that the time course of the change in the cellular components should coincide with the time course of the altered
S A Taran et al.
Prikladnaia biokhimiia i mikrobiologiia, 44(2), 181-186 (2008-08-02)
Cladribine (2-chloro-2'-deoxyadenosine) was synthesized using intact cells of the recombinant Escherichia coli strain producing Geobacillus stearothermophilus B-2194 thermostable purine-nucleoside phosphorylase II (EC 2.4.2.1). Use of the cells containing this thermostable enzyme allowed the process to be conducted at a temperature

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service