Skip to Content
Merck
All Photos(1)

Key Documents

Y0000599

Midazolam for system suitability

European Pharmacopoeia (EP) Reference Standard

Synonym(s):

Midazolam, 8-Chloro-6-(2-fluorophenyl)-1-methyl-4H-imidazo[1,5-a][1,4]benzodiazepine

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C18H13ClFN3
CAS Number:
Molecular Weight:
325.77
UNSPSC Code:
41116107
NACRES:
NA.24

grade

pharmaceutical primary standard

API family

midazolam

manufacturer/tradename

EDQM

drug control

regulated under CDSA - not available from Sigma-Aldrich Canada; psicótropo (Spain); Decreto Lei 15/93: Tabela IV (Portugal)

application(s)

pharmaceutical (small molecule)

format

neat

storage temp.

2-8°C

InChI

1S/C18H13ClFN3/c1-11-21-9-13-10-22-18(14-4-2-3-5-16(14)20)15-8-12(19)6-7-17(15)23(11)13/h2-9H,10H2,1H3

InChI key

DDLIGBOFAVUZHB-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the issuing Pharmacopoeia.For further information and support please go to the website of the issuing Pharmacopoeia.

Application

Midazolam for system suitability EP Reference standard, intended for use in laboratory tests only as specifically prescribed in the European Pharmacopoeia.

Packaging

The product is delivered as supplied by the issuing Pharmacopoeia. For the current unit quantity, please visit the EDQM reference substance catalogue.

Other Notes

Sales restrictions may apply.

Pictograms

Skull and crossbones

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Oral

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Eleonora L Swart et al.
Current drug metabolism, 13(6), 760-766 (2012-03-29)
A variety of developmental changes is of influence on the pharmacokinetics and pharmacodynamics of midazolam in neonatal and pediatric intensive care patients. However, dosing regimens in children are based upon rather empirical extrapolations from the dosing regimens in adults. Based
P Jevon
British dental journal, 213(2), 81-82 (2012-07-28)
This article is published in response to a query from reader Sarah Clements, based at Golding House Dental Practice Ltd in Kent, regarding the BDJ paper entitled 'Updated guidance on medical emergencies and resuscitation in the dental practice' (BDJ 2012;
Karly P Garnock-Jones
Paediatric drugs, 14(4), 251-261 (2012-06-19)
Oromucosal midazolam (Buccolam™) is a benzodiazepine approved for the treatment of pediatric patients with acute, prolonged, convulsive seizures. This article reviews the pharmacologic properties of oromucosal midazolam and its clinical efficacy and tolerability for the treatment of prolonged acute convulsive
Eugene Ng et al.
The Cochrane database of systematic reviews, 6(6), CD002052-CD002052 (2012-06-15)
Proper sedation for neonates undergoing uncomfortable procedures may reduce stress and avoid complications. Midazolam is a short-acting benzodiazepine that is increasingly used in neonatal intensive care units (NICU). However, its effectiveness as a sedative in neonates has not been systematically
Question 3 Ketamine or midazolam: does it matter which?
Elisabeth Jameson
Archives of disease in childhood, 96(1), 106-108 (2010-12-21)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service