Skip to Content
Merck
All Photos(1)

Key Documents

Y0000522

Norethisterone for system suitability

European Pharmacopoeia (EP) Reference Standard

Synonym(s):

19-Norethindrone, 17α-Ethynyl-19-nortestosterone, 17-Hydroxy-19-nor-17α-4-pregnen-20-yn-3-one, 17α-Ethynyl-17β-hydroxy-19-nor-4-androsten-3-one, 19-Nor-17α-ethynyl-4-androsten-17β-ol-3-one, 19-Norethisterone

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C20H26O2
CAS Number:
Molecular Weight:
298.42
Beilstein:
1915671
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

pharmaceutical primary standard

API family

norethisterone

manufacturer/tradename

EDQM

mp

205-206 °C (lit.)

application(s)

pharmaceutical (small molecule)

format

neat

storage temp.

−20°C

SMILES string

[H][C@]12CCC(=O)C=C1CC[C@]3([H])[C@]2([H])CC[C@@]4(C)[C@@]3([H])CC[C@@]4(O)C#C

InChI

1S/C20H26O2/c1-3-20(22)11-9-18-17-6-4-13-12-14(21)5-7-15(13)16(17)8-10-19(18,20)2/h1,12,15-18,22H,4-11H2,2H3/t15-,16+,17+,18-,19-,20-/m0/s1

InChI key

VIKNJXKGJWUCNN-XGXHKTLJSA-N

Gene Information

human ... PGR(5241)

Looking for similar products? Visit Product Comparison Guide

General description

This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the Issuing Pharmacopoeia. For further information and support please go to the website of the issuing Pharmacopoeia.

Application

Norethisterone for system suitability EP Reference standard, intended for use in laboratory tests only as specifically prescribed in the European Pharmacopoeia.

Biochem/physiol Actions

19-norethindrone is an oral contraceptive involved in the inhibition of cytosolic sulfotransferases (SULT).

Packaging

The product is delivered as supplied by the issuing Pharmacopoeia. For the current unit quantity, please visit the EDQM reference substance catalogue.

Other Notes

Sales restrictions may apply.

Pictograms

Health hazardEnvironment

Signal Word

Danger

Hazard Statements

Hazard Classifications

Aquatic Acute 1 - Aquatic Chronic 1 - Carc. 2 - Lact. - Repr. 1A

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Derin J Cobia et al.
Schizophrenia research, 139(1-3), 1-6 (2012-06-01)
Cortical abnormalities are considered a neurobiological characteristic of schizophrenia. However, the pattern of such deficits as they progress over the illness remains poorly understood. The goal of this project was to assess the progression of cortical thinning in frontal and
Patricia M LoRusso et al.
Cancer chemotherapy and pharmacology, 71(1), 193-202 (2012-10-16)
Vismodegib, a first-in-class oral hedgehog pathway inhibitor, is an effective treatment for advanced basal cell carcinoma. Based on in vitro data, a clinical drug-drug interaction (DDI) assessment of cytochrome P450 (CYP) 2C8 was necessary; vismodegib's teratogenic potential warranted a DDI
Chelsea B Polis et al.
The Lancet. Infectious diseases, 13(9), 797-808 (2013-07-23)
Whether or not the use of hormonal contraception affects risk of HIV acquisition is an important question for public health. We did a systematic review, searching PubMed and Embase, aiming to explore the possibility of an association between various forms
Kelli Stidham Hall et al.
Contraception, 86(6), 653-658 (2012-06-12)
Progestin-only contraceptive pills (POPs) offer a safe and effective contraceptive option, particularly for women at increased risk of venous thromboembolism. However, the prevalence of POP use among women in the United States is unknown. We analyzed population-based data from 12,279
Karl van den Broeck et al.
The European journal of contraception & reproductive health care : the official journal of the European Society of Contraception, 17(5), 321-328 (2012-09-15)
Ferdinand Peeters, a practising gynaecologist from Turnhout, near Antwerp (Belgium), had a pivotal role in the development of a practical and viable modality of oral contraception, striking the balance between efficacy and an acceptably low incidence and severity of side

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service