Skip to Content
Merck
All Photos(1)

Key Documents

Y0000511

Dexamethasone isonicotinate for impurity C identification

European Pharmacopoeia (EP) Reference Standard

Synonym(s):

Dexamethasone 21-isonicotinate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C28H32FNO6
CAS Number:
Molecular Weight:
497.56
UNSPSC Code:
41116107
NACRES:
NA.24

grade

pharmaceutical primary standard

API family

dexamethasone

manufacturer/tradename

EDQM

application(s)

pharmaceutical (small molecule)

format

neat

storage temp.

2-8°C

SMILES string

F[C@@]21[C@H]([C@H]4[C@@]([C@@]([C@@H](C4)C)(O)C(=O)COC(=O)c5ccncc5)(C[C@@H]2O)C)CCC3=CC(=O)C=C[C@@]31C

InChI

1S/C28H32FNO6/c1-16-12-21-20-5-4-18-13-19(31)6-9-25(18,2)27(20,29)22(32)14-26(21,3)28(16,35)23(33)15-36-24(34)17-7-10-30-11-8-17/h6-11,13,16,20-22,32,35H,4-5,12,14-15H2,1-3H3/t16-,20+,21+,22+,25+,26+,27+,28+/m1/s1

InChI key

BQTXJHAJMDGOFI-NJLPOHDGSA-N

General description

This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the issuing Pharmacopoeia.For further information and support please go to the website of the issuing Pharmacopoeia.

Application

Dexamethasone isonicotinate for impurity C identification EP Reference standard, intended for use in laboratory tests only as specifically prescribed in the European Pharmacopoeia.

Packaging

The product is delivered as supplied by the issuing Pharmacopoeia. For the current unit quantity, please visit the EDQM reference substance catalogue.

Other Notes

Sales restrictions may apply.

related product

Product No.
Description
Pricing

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Jitkamol Thanasak et al.
Veterinary research, 35(1), 103-112 (2004-04-22)
Dexamethasone is a potent therapeutic for treatment of the fatty liver syndrome or primary ketosis in post partum dairy cows. Reservations exist, however, among practitioners with respect to the risk of immunosuppression induced by corticosteroids. The aim of this study
G Hochhaus et al.
Journal of pharmaceutical and biomedical analysis, 9(9), 761-767 (1991-01-01)
A combined LC/RIA procedure is described for the selective determination of dexamethasone (DEX) and its prodrug dexamethasone-21-isonicotinate (DIN) in plasma. The low affinity of the employed dexamethasone antiserum for DIN (cross-reactivity less than 0.5%) allowed the direct determination of DEX
P L Toutain et al.
American journal of veterinary research, 45(9), 1750-1756 (1984-09-01)
Pharmacokinetics of dexamethasone and prednisolone were studied in 6 horses given dexamethasone alcohol (IV or IM) or dexamethasone 21-isonicotinate as a solution IV or IM (50 micrograms/kg of body weight), prednisolone 21-sodium succinate IV or IM (0.6 mg/kg of body
F T Cannizzo et al.
The Veterinary record, 163(15), 448-452 (2008-10-14)
Three groups of 10 veal calves were treated, respectively, with 5 mg of dexamethasone-21-isonicotinate administered intramuscularly on days 0 and 7 (group A); 0.4 mg/day of dexamethasone-21-phosphate administered orally for 20 days (group B); or left untreated as controls (group
[Modern aerosols in the treatment of obstructive respiratory tract diseases. 3. Inhalable corticoids].
D Heide
Zeitschrift fur arztliche Fortbildung, 78(2), 49-51 (1984-01-01)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service