Skip to Content
Merck
All Photos(1)

Key Documents

Y0000490

Misoprostol

European Pharmacopoeia (EP) Reference Standard

Synonym(s):

(±)-15-Deoxy-(16RS)-16-hydroxy-16-methylprostaglandin E1 methyl ester

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C22H38O5
CAS Number:
Molecular Weight:
382.53
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

pharmaceutical primary standard

API family

misoprostol

manufacturer/tradename

EDQM

application(s)

pharmaceutical (small molecule)

format

neat

storage temp.

−20°C

SMILES string

CCCCC(C)(O)C\C=C\[C@H]1[C@H](O)CC(=O)[C@@H]1CCCCCCC(=O)OC

InChI

1S/C22H38O5/c1-4-5-14-22(2,26)15-10-12-18-17(19(23)16-20(18)24)11-8-6-7-9-13-21(25)27-3/h10,12,17-18,20,24,26H,4-9,11,13-16H2,1-3H3/b12-10+/t17-,18-,20-,22?/m1/s1

InChI key

OJLOPKGSLYJEMD-URPKTTJQSA-N

Gene Information

human ... PTGER3(5733)

Looking for similar products? Visit Product Comparison Guide

General description

This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the Issuing Pharmacopoeia. For further information and support please go to the website of the issuing Pharmacopoeia.

Application

Misoprostol EP Reference standard, intended for use in laboratory tests only as specifically prescribed in the European Pharmacopoeia.

Biochem/physiol Actions

PGE1 analog prodrug which is rapidly de-esterified to active "misoprostolic acid". Cited for extremely wide-ranging therapeutic effects, including prevention of NSAID-induced gastric ulceration, regulation of immunologic cascades, inhibition of platelet-activating factor (PAF), treatment of ethanol- and acetaminophen-induced hepatotoxicity and hepatitis, and stimulation of cartilage repair after injury.

Packaging

The product is delivered as supplied by the issuing Pharmacopoeia. For the current unit quantity, please visit the EDQM reference substance catalogue.

Other Notes

Sales restrictions may apply.

Pictograms

Skull and crossbonesHealth hazard

Signal Word

Danger

Hazard Statements

Precautionary Statements

Hazard Classifications

Acute Tox. 3 Oral - Repr. 1B

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Christina S Chu et al.
Journal of the Royal Society of Medicine, 105(8), 336-347 (2012-08-22)
This article describes and critically appraises clinical trials assessing misoprostol effectiveness in preventing primary postpartum haemorrhage (PPH) in home and community settings in low- and middle-income countries. Of 172 identified studies of misoprostol use in labour only six fulfilled the
Jeanine F Carbone et al.
Obstetrics and gynecology, 121(2 Pt 1), 247-252 (2013-01-11)
To test the hypothesis that use of the Foley bulb plus vaginal misoprostol will result in shorter induction-to-delivery time compared with vaginal misoprostol alone. We randomized 123 women undergoing induction of labor with singleton pregnancies at 24 weeks of gestation
V A Hundley et al.
BJOG : an international journal of obstetrics and gynaecology, 120(3), 277-285 (2012-11-30)
Using misoprostol to prevent postpartum haemorrhage (PPH) in home-birth settings remains controversial. To review the safety and effectiveness of oral misoprostol in preventing PPH in home-birth settings. The Cochrane Library, PubMed, and POPLINE were searched for articles published until 31
J Hua et al.
BJOG : an international journal of obstetrics and gynaecology, 120(5), 531-540 (2013-01-22)
The efficacy of misoprostol versus oxytocin for reducing blood loss during caesarean section remains unclear. To conduct a meta-analysis comparing the efficacy of misoprostol with that of oxytocin in reducing blood loss during caesarean section. We searched MEDLINE, Embase, the
Amanda Selk et al.
Obstetrics and gynecology, 118(4), 941-949 (2011-09-22)
To estimate the benefits and harms of misoprostol use for cervical dilation in patients undergoing operative hysteroscopy. We searched MEDLINE, EMBASE, and the Cochrane Central Register of Controlled Trials (from inception to February 2011). We also searched trial registries, other

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service