Skip to Content
Merck
All Photos(1)

Key Documents

Y0000354

Acarbose for identification

European Pharmacopoeia (EP) Reference Standard

Synonym(s):

Acarbose, 4",6"-Dideoxy-4"-([1S]-[1,4,6/5]-4,5,6-trihydroxy-3-hydroxymethyl-2-yclohexenylamino)-maltotriose

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C25H43NO18
CAS Number:
Molecular Weight:
645.60
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

pharmaceutical primary standard

API family

acarbose

manufacturer/tradename

EDQM

application(s)

pharmaceutical (small molecule)

format

neat

storage temp.

2-8°C

SMILES string

C[C@H]1O[C@H](O[C@H]2[C@H](O)[C@@H](O)[C@H](O[C@@H]2CO)O[C@H]3[C@H](O)[C@@H](O)[C@H](O)O[C@@H]3CO)[C@H](O)[C@@H](O)[C@@H]1N[C@H]4C=C(CO)[C@@H](O)[C@H](O)[C@H]4O

InChI

1S/C25H43NO18/c1-6-11(26-8-2-7(3-27)12(30)15(33)13(8)31)14(32)19(37)24(40-6)43-22-10(5-29)42-25(20(38)17(22)35)44-21-9(4-28)41-23(39)18(36)16(21)34/h2,6,8-39H,3-5H2,1H3/t6-,8+,9-,10-,11-,12-,13+,14+,15+,16-,17-,18-,19-,20-,21-,22-,23-,24-,25-/m1/s1

InChI key

XUFXOAAUWZOOIT-SXARVLRPSA-N

Gene Information

Looking for similar products? Visit Product Comparison Guide

General description

This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the issuing Pharmacopoeia.For further information and support please go to the website of the issuing Pharmacopoeia.

Application

Acarbose for identification EP Reference standard, intended for use in laboratory tests only as specifically prescribed in the European Pharmacopoeia.

Biochem/physiol Actions

Modified tetrasaccharide that acts as a reversible α -glucosidase inhibitor.

Packaging

The product is delivered as supplied by the issuing Pharmacopoeia. For the current unit quantity, please visit the EDQM reference substance catalogue.

Other Notes

Sales restrictions may apply.

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Sophie Delorme et al.
Current opinion in pharmacology, 5(2), 184-189 (2005-03-23)
The prevalence of glucose intolerance is increasing dramatically worldwide. Both impaired glucose tolerance (IGT) and type 2 diabetes are associated with excess mortality from cardiovascular disease. It is now generally accepted that these cardiovascular complications are related to prevailing hyperglycemia
L Furio et al.
Minerva gastroenterologica e dietologica, 52(3), 339-346 (2006-09-15)
The authors report a case report of rare disease interesting the digestive tract and often associated to the other gastrointestinal pathologies and/or pulmonary diseases and can be also associated to not gastrointestinal conditions such as collagen-vascular disease, transplantation, AIDS, use
Peter N Båvenholm et al.
Diabetes & vascular disease research, 3(2), 72-79 (2006-10-25)
Traditional risk factors do not fully explain the increased risk of cardiovascular disease (CVD) in diabetes. Epidemiology shows that hyperglycaemia is a continuous CVD risk factor and that two-hour postprandial glucose levels are more strongly associated with CVD than fasting
Matteo Monami et al.
Advances in therapy, 29(9), 736-746 (2012-08-28)
Lipid profile is an important determinant of cardiovascular risk in type 2 diabetes. It is well known that patients with type 2 diabetes are more likely to be dyslipidemic than the general population. Given the observed connection between glucose and
J de la Vega et al.
Gastroenterologia y hepatologia, 23(6), 282-284 (2004-08-25)
A 57-year-old woman with non-insulin-dependent diabetes mellitus and inadequate glycemic control was prescribed acarbose (100 mg 3 times daily). Two months later she presented with acute hepatitis (ALT 2,300 IU/l). Other causes of liver damage were excluded. Three months after

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service